Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Globins are a superfamily of gas-binding heme proteins that are present in bacteria, protists, fungi, plants and animals. Globins play evolutionarily divergent roles which include binding, transport, scavenging, detoxification, and sensing of oxygen, nitric oxide and carbon monoxide. Neuroglobin (Ngb) is a hexacoordinate hemoglobin that is predominantly expressed in the vertebrate brain and may enhance oxygen supply to neural components. Neuroglobin displays a high affinity for oxygen and its presence in cerebral neurons suggests a role in neuronal responses to hypoxia or ischemia. For example, in vitro neuronal hypoxia causes an elevation in the levels of neuroglobin, which enhances neuronal cell survival. The human neuroglobin gene maps to chromosome 14q24 and encodes a 151 amino acid protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuroglobin; Nitrite reductase
Gene Aliases: NGB
UniProt ID: (Human) Q9NPG2, (Rat) Q99JA8, (Mouse) Q9ER97
Entrez Gene ID: (Human) 58157, (Rat) 85382, (Mouse) 64242
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support