Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Electroneutral exchange of protons for Na(+) and Li(+) across the inner mitochondrial membrane. Contributes to the organellar volume homeostasis. Required for osteoclast differentiation and bone resorption activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mitochondrial Na(+)/H(+) exchanger NHA2; mitochondrial sodium/hydrogen exchanger 9B2; mitochondrial sodium/hydrogen exchanger NHA2; Na(+)/H(+) exchanger NHA2; Na(+)/H(+) exchanger-like domain-containing protein 2; Na+/H+ exchanger domain containing 2; NHA-oc; NHE domain-containing protein 2; Soditlum/hydrogen exchanger-like domain-containing protein 2; Sodium/hydrogen exchanger 9B2; Sodium/hydrogen exchanger-like domain-containing protein 2; Solute carrier family 9 subfamily B member 2; solute carrier family 9, subfamily B (cation proton antiporter 2), member 2; solute carrier family 9, subfamily B (NHA2, cation proton antiporter 2), member 2
Gene Aliases: C80638; nha-oc; NHA2; nhaoc; NHE10; NHEDC2; SLC9B2
UniProt ID: (Human) Q86UD5, (Mouse) Q5BKR2
Entrez Gene ID: (Human) 133308, (Mouse) 97086, (Rat) 679958
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support