Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SDLEPVEEAEGDGKGGSRAALYPHPQQLSREEKRRRRRATAKYRSAHATR ERIRVEAFNLAFAELRKLLPTLPPDKKLSKIEILRLAICYISYLNHVLDV
May serve as DNA-binding protein and may be involved in the control of cell-type determination, possibly within the developing nervous system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa34; Class A basic helix-loop-helix protein 34; Helix-loop-helix protein 2; HEN-2; Nescient helix loop helix 2; NSCL-2
Gene Aliases: BHLHA34; HEN2; KIAA0490; NHLH2; NSCL2
UniProt ID: (Human) Q02577
Entrez Gene ID: (Human) 4808
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support