Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MLASSSRIRA AWTRALLLPL LLAGPVGCLS RQELFPFGPG QGDLELEDGD DFVSPALELR GALRFYDRSD IDAVYVTTNG IIATSEPPAK ESHPGLFPPT FGAVAPFLAD LDTTDGLGKV YYREDLSPSI TQRAAECVHR GFPEISFQPS SAVVVTWESV APYQGPSRDP DQKGKRNTFQ AVLASSDSSS YAIFLYPEDG LQFHTTFSKK ENNQVPAVVA FSQGSVGFLW KSNGAYNIFA NDRESIENLA KSSNSGQQGV WVFEIGSPAT TNGVVPADVI LGTEDGAEYD DEDEDYDLAT TRLGLEDVGT TPFSYKALRR GGADTYSVPS VLSPRRAATE RPLGPPTERT RSFQLAVETF HQQHPQVIDV DEVEETGVVF SYNTDSRQTC ANNRHQCSVH AECRDYATGF CCSCVAGYTG NGRQCVAEGS PQRVNGKVKG RIFVGSSQVP IVFENTDLHS YVVMNHGRSY TAISTIPETV GYSLLPLAPV GGIIGWMFAV EQDGFKNGFS ITGGEFTRQA EVTFVGHPGN LVIKQRFSGI DEHGHLTIDT ELEGRVPQIP FGSSVHIEPY TELYHYSTSV ITSSSTREYT VTEPERDGAS PSRIYTYQWR QTITFQECVH DDSRPALPST QQLSVDSVFV LYNQEEKILR YALSNSIGPV REGSPDALQN PCYIGTHGCD TNAACRPGPR TQFTCECSIG FRGDGRTCYE VEKTRCQHER EHILGAAGAT DPQRPIPPGL FVPECDAHGH YAPTQCHGST GYCWCVDRDG REVEGTRTRP GMTPPCLSTV APPIHQGPAV PTAVIPLPPG THLLFAQTGK IERLPLEGNT MRKTEAKAFL HVPAKVIIGL AFDCVDKMVY WTDITEPSIG RASLHGGEPT TIIRQDLGSP EGIAVDHLGR NIFWTDSNLD RIEVAKLDGT QRRVLFETDL VNPRGIVTDS VRGNLYWTDW NRDNPKIETS YMDGTNRRIL VQDDLGLPNG LTFDAFSSQL CWVDAGTNRA ECLNPSQPSR RKALEGLQYP FAVTSYGKNL YFTDWKMNSV VALDLAISKE TDAFQPHKQT RLYGITTALS QCPQGHNYCS VNNGGCTHLC LATPGSRTCR CPDNTLGVDC IEQK
This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: enactin; Entactin; entactin 1; NID 1; NID-1; Nidogen (enactin); Nidogen precursor (Entactin); Nidogen-1; Nidogen-1;
Gene Aliases: A630025O17; Ent; entactin; entactin-1; NID; NID1; nidogen-1
UniProt ID: (Human) P14543, (Mouse) P10493
Entrez Gene ID: (Human) 4811, (Mouse) 18073
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support