Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Has a omega-amidase activity. The role of omega-amidase is to remove potentially toxic intermediates by converting alpha-ketoglutaramate and alpha-ketosuccinamate to biologically useful alpha-ketoglutarate and oxaloacetate, respectively. Overexpression decreases the colony-forming capacity of cultured cells by arresting cells in the G2 phase of the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory sperm binding protein Li 8a; Nit protein 2; nitrilase family, member 2; Nitrilase homolog 2; Omega-amidase NIT2
Gene Aliases: CUA002; HEL-S-8a; NIT2
UniProt ID: (Human) Q9NQR4
Entrez Gene ID: (Human) 56954
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support