Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGKLQSKHA AAARKRRESP EGDSFVASAY ASGRKGAEEA ERRARDKQEL PNGDPKEGPF REDQCPLQVA LPAEKAEGRE HPGQLLSADD GERAANREGP RGPGGQRLNI DALQCDVSVE EDDRQEWTFT LYDFDNCGKV TREDMSSLMH TIYEVVDASV NHSSGSSKTL RVKLTVSPEP SSKRKEGPPA GQDREPTRCR MEGELAEEPR VADRRLSAHV RRPSTDPQPC SERGPYCVDE NTERRNHYLD LAGIENYTSR FGPGSQLCEK RSSAPRTHSG DKARGVGLCR ELWSQAGHPQ WPGPFPSGLV AV (1-311 aa encoded by BC012176)
Cell autonomous antagonist of the canonical Wnt signaling pathway. May activate a second Wnt signaling pathway that controls planar cell polarity. Required for processing of TGFA and for targeting of TGFA to the basolateral membrane of polarized epithelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dvl-binding protein NKD2; hNkd2; mNkd2; naked cuticle 1 homolog; naked cuticle 2 homolog; naked cuticle-2; Naked-2; Protein naked cuticle homolog 2
Gene Aliases: 2210403L10Rik; AW212591; Naked2; NKD2
UniProt ID: (Human) Q969F2, (Mouse) Q8VE28
Entrez Gene ID: (Human) 85409, (Rat) 308068, (Mouse) 72293
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support