Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDNQGVIYS DLNLPPNPKR QQRKPKGNKS SILATEQEIT YAELNLQKAS QDFQGNDKTY HCKDLPSAPE KLIVGILGII CLILMASVVT IVVIPSRHCG HCPEEWITYS NSCYYIGKER RTWEESLLAC TSKNSSLLSI DNEEEMKFLS IISPSSWIGV FRNSSHHPWV TMNGLAFKHE IKDSDNAELN CAVLQVNRLK SAQCGSSIIY HCKHKL (1-215 aa encoded by B C012550)
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. The protein encoded by this gene belongs to the killer cell lectin-like receptor family, also called NKG2 family, which is a group of transmembrane proteins preferentially expressed in NK cells. This family of proteins is characterized by the type II membrane orientation and the presence of a C-type lectin domain. This protein forms a complex with another family member, KLRD1/CD94, and has been implicated in the recognition of the MHC class I HLA-E molecules in NK cells. The genes of NKG2 family members form a killer cell lectin-like receptor gene cluster on chromosome 12. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-lectin type II protein; CD159 antigen-like family member A; CD159a; killer cell lectin-like receptor subfamily C, member 1; natural killer cell lectin; natural killer group protein 2; NK cell receptor A; NKG2-1/B activating NK receptor; NKG2-A/B type II integral membrane protein; NKG2-A/B-activating NK receptor; NKG2-A/NKG2-B type II integral membrane protein
Gene Aliases: CD159A; KLRC1; NKG2; NKG2A
UniProt ID: (Human) P26715
Entrez Gene ID: (Human) 3821
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support