Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CAVLQVNRLKS AQCGSSMIYH CKH
Highest antigen sequence identity to the following orthologs - mouse 38%, rat 38%.
Natural killer (NK) cells are lymphocytes that can mediate lysis of certain tumor cells and virus-infected cells without previous activation. They can also regulate specific humoral and cell-mediated immunity. The protein encoded by this gene belongs to the killer cell lectin-like receptor family, also called NKG2 family, which is a group of transmembrane proteins preferentially expressed in NK cells. This family of proteins is characterized by the type II membrane orientation and the presence of a C-type lectin domain. This protein forms a complex with another family member, KLRD1/CD94, and has been implicated in the recognition of the MHC class I HLA-E molecules in NK cells. The genes of NKG2 family members form a killer cell lectin-like receptor gene cluster on chromosome 12. Four alternatively spliced transcript variants encoding two distinct isoforms have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD159 antigen-like family member C; CD159A; CD159c; cd159e; killer cell lectin-like receptor subfamily C, member 2; KLRC1; NK cell receptor C; NKG2; NKG2-C type II integral membrane protein; NKG2-C-activating NK receptor; NKG2A
Gene Aliases: CD159c; KLRC2; NKG2-C; NKG2C
UniProt ID: (Human) P26717
Entrez Gene ID: (Human) 3822
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support