Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
KappaB ras-1 (kappa-B-ras-1) and kappaB-ras-2 are two small proteins that similar to Ras-like small GTPases that associate with Ikappa-B (Ikappa-B), an inhibitor of the transcription factor NF-kappa-B. Ikappa-B exists in two homologous forms, Ikappa-B-alpha and Ikappa-B-beta, although Ikappa-B-beta contains a unique 47-amino acid region within its ankyrin domain. While inactive Ikappa-B-alpha-NF-kappa-B complexes can shuttle in and out of the nucleus, Ikappa-B-beta-NF-kappa-B complexes are retained exclusively in the cytoplasm. It is suggested that kappaB-ras proteins preferentially bind to the Ikappa-B-beta form through this unique insert within the ankyrin region, thus modulating the cellular location of Ikappa-B-beta and regulating the rate of degradation of Ikappa-B-beta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C15orf1; I-kappa-B-interacting Ras-like protein 1; I1PP2A; kappa B-ras 1; Kappa B-Ras protein 1; MAPM; MGC119787; MGC150373; NF-kappa-B inhibitor-interacting Ras-like protein 1; NFKB inhibitor interacting Ras-like 1; NFKB inhibitor interacting Ras-like protein 1; PHAP1; PP32
Gene Aliases: kappaB-Ras1; KBRAS1; NKIRAS1
UniProt ID: (Human) Q9NYS0
Entrez Gene ID: (Human) 28512
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support