Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGKSCKVVV CGQASVGKTS ILEQLLYGNH VVGSEMIETQ EDIYVGSIET DRGVREQVRF YDTRGLRDGA ELPRHCFSCT DGYVLVYSTD SRESFQRVEL LKKEIDKSKD KKEVTIVVLG NKCDLQEQRR VDPDVAQHWA KSEKVKLWEV SVADRRSLLE PFVYLASKMT QPQSKSAFPL SRKNKGSGSL DG (1-191 aa encoded by BC007450)
Atypical Ras-like protein that acts as a potent regulator of NF-kappa-B activity by preventing the degradation of NF-kappa-B inhibitor beta (NFKBIB) by most signals, explaining why NFKBIB is more resistant to degradation. May act by blocking phosphorylation of NFKBIB and nuclear localization of p65/RELA NF-kappa-B subunit. It is unclear whether it acts as a GTPase. Both GTP- and GDP-bound forms block phosphorylation of NFKBIB.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: I-kappa-B-interacting Ras-like protein 2; Kappa B-Ras protein 2; NF-kappa-B inhibitor-interacting Ras-like protein 2; NFKB inhibitor interacting Ras-like 2; NFKB inhibitor interacting Ras-like protein 2
Gene Aliases: kappaB-Ras2; KBRAS2; NKIRAS2
UniProt ID: (Human) Q9NYR9
Entrez Gene ID: (Human) 28511
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support