Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the NLR family. Alternative splicing has been observed at this gene locus and two transcript variants, encoding distinct isoforms, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Caterpiller protein 11.3; CLR11.3; FLJ21478; MGC131937; MGC21025; NLR family member X1; NOD-like receptor X1; nucleotide-binding oligomerization domain protein 26; Nucleotide-binding oligomerization domain protein 5; Nucleotide-binding oligomerization domain protein 9; nucleotide-binding oligomerization domain, leucine rich repeat containing X1; OTTHUMP00000205768; OTTHUMP00000205769
Gene Aliases: CLR11.3; DLNB26; NLRX1; NOD26; NOD5; NOD9
UniProt ID: (Human) Q86UT6
Entrez Gene ID: (Human) 79671
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support