Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide shares 95% identity with mouse sequence.
Specificity of this antibody: NMDAR3A.
Grin3a is an NMDA (N-methyl-D-aspartate) receptor subtype of glutamate-gated ion channels with reduced single-channel conductance, low calcium permeability and low voltage-dependent sensitivity to magnesium. Grin3a is mediated by glycine and may play a role in the development of dendritic spines. Further, Grin3a may play a role in PPP2CB-NMDAR mediated signaling mechanism. Grin3a belongs to the superfamily of glutamate-regulated ion channels, and function in physiological and pathological processes in the central nervous system. Grin3a shows greater than 90% identity to the corresponding subunit in rat. Studies in the knockout mouse deficient in Grin3a suggest that the protein may be involved in the development of synaptic elements by modulating NMDA receptor activity. Diseases associated with GRIN3A include Lingual-Facial-Buccal Dyskinesia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GluN3A; glutamate receptor; Glutamate receptor chi-1; Glutamate receptor ionotropic, NMDA 3A; N-methyl-D-aspartate receptor; N-methyl-D-aspartate receptor subtype 3A; NMDAR-L; NMDAR-L1; NMDAR3A
Gene Aliases: chi-1; GluN3A; Grin3a; NR3
UniProt ID: (Rat) Q9R1M7
Entrez Gene ID: (Rat) 191573
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support