Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEIQELEEIQ ACQGLWEVFV TLSERARDYL HKTGRFIVIG GIVSPVHDSY GKQGLVSSRH RLIMCQLAVQ NSDWIRVDPW ECYQDTWQTT CSVLEHHRDL MKRVTGCILS NVNTPSMTPV IGQPQNETPQ PIYQNSNVAT KPTAAKILGK VGESLSRICC VRPPVERFTF VDENANLGTV MRYEEIELRI LLLCGSDLLE SFCIPGLWNE ADMEVIVGDF GIVVVPRDAA DTDRIMNHSS ILRKYKNNIM VVKDDINHPM SVVSSTKSRL ALQHGDGHVV DYLSQPVIDY ILKSQLYINA SG
Catalyzes the formation of NAD(+) from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form; nicotinic acid mononucleotide (NaMN) as substrate but with a lower efficiency. Cannot use triazofurin monophosphate (TrMP) as substrate. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD(+). For the pyrophosphorolytic activity prefers NAD(+), NADH and NAAD as substrates and degrades nicotinic acid adenine dinucleotide phosphate (NHD) less effectively. Fails to cleave phosphorylated dinucleotides NADP(+), NADPH and NAADP(+).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NaMN adenylyltransferase 2; Nicotinamide mononucleotide adenylyltransferase 2; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 2; Nicotinate-nucleotide adenylyltransferase 2; NMN adenylyltransferase 2; NMN/NaMN adenylyltransferase 2; pyridine nucleotide adenylyltransferase 2
Gene Aliases: C1orf15; KIAA0479; NMNAT2; PNAT2
UniProt ID: (Human) Q9BZQ4
Entrez Gene ID: (Human) 23057
Molecular Function:
nucleotidyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support