Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: CIPGLWNEAD MEVIVGDFGI VVVPRDAADT DRIMNHSSIL RKYKNNIMVV KDDINHPMSV VSSTKSRLAL QHGDGHVVDY LSQPVIDYIL KSQLYINASG
Catalyzes the formation of NAD(+) from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form; nicotinic acid mononucleotide (NaMN) as substrate but with a lower efficiency. Cannot use triazofurin monophosphate (TrMP) as substrate. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD(+). For the pyrophosphorolytic activity prefers NAD(+), NADH and NAAD as substrates and degrades nicotinic acid adenine dinucleotide phosphate (NHD) less effectively. Fails to cleave phosphorylated dinucleotides NADP(+), NADPH and NAADP(+).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NaMN adenylyltransferase 2; Nicotinamide mononucleotide adenylyltransferase 2; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 2; Nicotinate-nucleotide adenylyltransferase 2; NMN adenylyltransferase 2; NMN/NaMN adenylyltransferase 2; pyridine nucleotide adenylyltransferase 2
Gene Aliases: C1orf15; KIAA0479; NMNAT2; PNAT2
UniProt ID: (Human) Q9BZQ4
Entrez Gene ID: (Human) 23057
Molecular Function:
nucleotidyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support