Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNRSLANVIL GGYGTTSTAG GKPMEISGTH TEINLDNAID MIREANSIII TPGYGLCAAK AQYPIADLVK MLTEQGKKVR FGIHPVAGRM PGQLNVLLAE AGVPYDIVLE MDEINHDFPD TDLVLVIGAN DTVNSAAQED PNSIIAGMPV LEVWKSKQVI VMKRSLGVGY AAVDNPIFYK PNTAMLLGDA KKTCDALQAK VRESYQK
This gene encodes an integral protein of the inner mitochondrial membrane. The enzyme couples hydride transfer between NAD(H) and NADP(+) to proton translocation across the inner mitochondrial membrane. Under most physiological conditions, the enzyme uses energy from the mitochondrial proton gradient to produce high concentrations of NADPH. The resulting NADPH is used for biosynthesis and in free radical detoxification.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NAD(P) transhydrogenase, mitochondrial; Nicotinamide nucleotide transhydrogenase; Nicotinamide nucleotide transhydrogenase (NAD(P)+ transhydrogenase); Pyridine nucleotide transhydrogenase
Gene Aliases: 4930423F13Rik; AI323702; BB168308; GCCD4; NNT
UniProt ID: (Human) Q13423
Entrez Gene ID: (Rabbit) 100341011, (Human) 23530, (Pig) 100526144, (Rat) 310378, (Mouse) 18115
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support