Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MADAGIRRVV PSDLYPLVLG FLRDNQLSEV ANKFAKATGA TQQDANASSL LDIYSFWLNR SAKVPERKLQ ANGPVAKKAK KKASSSDSED SSEEEEEVQG PPAKKAAVPA KRVGLPPGKA AAKASESSSS EESSDDDDEE DQKKQPVQKG VKPQAKAAKA PPKKAKSSDS DSDSSSEDEP PKNQKPKITP VTVKAQTKAP PKPARAAPKI ANGKAASSSS SSSSSSSSDD SEEEKAAATP KKTVPKKQVV AKAPVKAATT PTRKSSSSED SSSDEEEEQK KPMKNKPGPY SSVPPPSAPP PKKSLGTQPP KKAVEKQQPV ESSEDSSDES DSSSEEEKKP PTKAVVSKAT TKPPPAKKAA ESSSDSSDSD SSEDDEAPSK PAGTTKNSSN KPAVTTKSPA VKPAAAPKQP VGGGQKLLTR KADSSSSEEE SSSSEEEKTK KMVATTKPKA TAKAALSLPA KQAPQGSRDS SSDSDSSSSE EEEEKTSKSA VKKKPQKVAG GAAPSKPASA KKGKAESSNS SSSDDSSEEE EEKLKGKGSP RPQAPKANGT SALTAQNGKA AKNSEEEEEE KKKAAVVVSK SGSLKKRKQN EAAKEAETPQ AKKIKLQTPN TFPKRKKGEK RASSPFRRVR EEEIEVDSRV ADNSFDAKRG AAGDWGERAN QVLKFTKGKS FRHEKTKKKR GSYRGGSISV QVNSIKFDSE
Related to nucleologenesis, may play a role in the maintenance of the fundamental structure of the fibrillar center and dense fibrillar component in the nucleolus. It has intrinsic GTPase and ATPase activities. May play an important role in transcription catalyzed by RNA polymerase I.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 140 kDa nucleolar phosphoprotein; HCV NS5A trans-regulated protein 13; HCV NS5A-transactivated protein 13; Hepatitis C virus NS5A-transactivated protein 13; Nopp140; Nucleolar 130 kDa protein; Nucleolar and coiled-body phosphoprotein 1; nucleolar and coiled-body phosphprotein 1; Nucleolar phosphoprotein p130; nucleolar protein p130
Gene Aliases: KIAA0035; NOLC1; NOPP130; NOPP140; NS5ATP13; P130
UniProt ID: (Human) Q14978
Entrez Gene ID: (Human) 9221
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support