Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Involved in nucleolar processing of pre-18S ribosomal RNA. Has a role in the nuclear export of 40S pre-ribosomal subunit to the cytoplasm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NOP14 nucleolar protein homolog; Nucleolar complex protein 14; Nucleolar protein 14; probable nucleolar complex protein 14
Gene Aliases: C4orf9; NOL14; NOP14; RES4-25; RES425; UTP2
UniProt ID: (Human) P78316
Entrez Gene ID: (Human) 8602
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support