Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PEQPFEKAAF QKQNDTPKGP QPPTVSPIRS SRPPPAKRKK SQSRGNSQLL
For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
NOP2 is a member of the unc-84 homolog family and encodes a nuclear nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S rRNA (cytosine(4447)-C(5))-methyltransferase; MGC117384; MGC149287; MGC149288; NOL1/NOP2/Sun domain family, member 1; NOP2 nucleolar protein homolog; nop2p; Nucleolar protein 1; nucleolar protein 1, 120kDa; Nucleolar protein 2 homolog; Proliferating-cell nucleolar antigen p120; Proliferation-associated nucleolar protein p120; putative ribosomal RNA methyltransferase NOP2
Gene Aliases: NOL1; NOP120; NOP2; NSUN1; p120
UniProt ID: (Human) P46087
Entrez Gene ID: (Human) 4839
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support