Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MMAIAPNVT VMVGELVGAR LIAHAGSLLN LAKHAASTVQ ILGAEKALFR ALKSRRDTPK YGLIYHASLV GQTSPKHKGK ISRMLAAKTV LAIRYDAFGE DSSSAMGVEN RAKLEARLRT LEDRGIRKIS GTGKALAKTE KYEHKSEVKT YDPSGDSTLP TCSKKRKIEQ VDKEDEITEK KAKKAKIKVK VEEEEEEKVA EEEETSVKKK KKRGKKKHIK EEPLSEEEPC TSTAIASPEK KKKKKKKREN ED (279-529 aa encoded by BC032592)
Nop5p, or Nop58p, is localized in the nucleolus and required for pre-18S rRNA processing in Saccharomyces cerevisiae.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MSSP; NOP58 ribonucleoprotein homolog; Nopp140 associated protein; Nopp140-associated protein of 65 kDa; Nucleolar protein 5; Nucleolar protein 58; nucleolar protein NOP5/NOP58; nucleolar protein NOP58; SIK similar protein; SIK-similar protein
Gene Aliases: HSPC120; MSSP; Nap65; NOL5; NOP5; NOP5/NOP58; NOP58; SIK
UniProt ID: (Human) Q9Y2X3, (Mouse) Q6DFW4, (Rat) Q9QZ86
Entrez Gene ID: (Human) 51602, (Mouse) 55989, (Rat) 60373
Molecular Function: RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support