Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: SEGRKTWRRR GQQPPPPPRT EAAPAAGQPV ESFPLDFTAV EGNMDSFMAQ VKSLAQSLYP CSAQQLNEDL RLHLLLNTSV TCNDGSPAGY YLKESRGSRR WLLFLEGGWY CFNRENCDSR YDTMRRLMSS RDWPRTRTGT GILSSQPEEN PYWWNANMVF IPYCSSDVWS GASSKSEKNE YAFMGALIIQ EVVRELLGRG LSGAKVLLLA GSSAGGTGVL LNVDRVAEQL EKLGYPAIQV RGLADSGWFL DNKQYRHTDC VDTITCAPTE AIRRGIRYWN GVVPERCRRQ FQEGEEWNCF F; Positive Samples: BxPC-3, HepG2
Genetic studies in Drosophila have revealed a novel secreted antagonist for Wingless (Wg) signaling, referred to as Notum/Wingful (Wf). Wf/Notum encodes a protein of 430 amino acid residues, which is related to plant pectin acetylesterases of the a/beta-hydrolase superfamily. The main target of Notum action appears to be cell surface proteoglycans Dally and/or Dally-like (Dlp). Notum governs the extracellular Wg morphogen distribution via Dally and Dlp. Also, like some vertebrate Wnt antagonists, notum expression is activated by Wg signaling, revealing the existence of a feedback control mechanism in which Wg modifies its own gradient distribution. In vertebrate, proteoglycans related to Dally and Dlp likely play important roles in Wnt signaling which encodes a glypican that is required for Wnt-11 signaling. A single ortholog of Notum exists in the mouse and the human genome, implying that a similar regulatory mechanism may exist in vertebrates.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: [Wnt protein] O-palmitoleoyl-L-serine hydrolase; hNOTUM; notum pectinacetylesterase homolog; Palmitoleoyl-protein carboxylesterase NOTUM; protein notum homolog
Gene Aliases: hNOTUM; NOTUM; OK/SW-CL.30
UniProt ID: (Human) Q6P988
Entrez Gene ID: (Human) 147111
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support