Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MQSVQSTSFC LRKQCLCLTF LLLHLLGQVA ATQRCPPQCP GRCPATPPTC APGVRAVLDG CSCCLVCARQ RGESCSDLEP CDESSGLYCD RSADPSKQTG ICTAVEGDNC VFDGVIYRSG EKFQPSCKFQ CTCRDGQIGC VPRCQLDVLL PEPNCPAPRK VEVPGECCEK WICGPDEEDS LGGLTLAAYR PEATLGVEVS DSSVNCIEQT TEWTACSKSC GMGFSTRVTN RNRQCEMLKQ TRLCMVRPCE QEPEQPTDKK GKKCLRTKKS LKAIHLQFKN CTSLHTYKPR FCGVCSDGRC CTPHNTKTIQ AEFQCSPGQI VKKPVMVIGT CTCHTNCPKN NEAFLQELEL KTTRGKM
NOV is a member of the CCN family of secreted, cysteine-rich regulatory proteins. The full-length NOV protein contains four structural domains that confer distinct, and sometimes opposing, biological activities. Elevated expression of NOV is associated with certain tumors, including Wilm's tumor and most nephroblastomas. However, in other tumor types and certain cancer cell lines, increased tumorigenicity and proliferation is correlated with decreased NOV expression. Additionally, NOV induces cell adhesion and cell migration by signaling through specific cell-surface integrins, and by binding to heparin sulfate proteoglycans and to fibulin 1C. NOV has also been reported to exert proangiogenic activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCN family member 3; Cellular communication network factor 3; IBP-9; IGF-binding protein 9; Insulin-like growth factor-binding protein 9; Nephro blastoma-overexpressed gene protein homolog; nephroblastoma-overexpressed gene protein homolog; NovH; Protein NOV homolog
Gene Aliases: CCN3; IBP-9; IGFBP-9; IGFBP9; NOV; NOVH
UniProt ID: (Human) P48745
Entrez Gene ID: (Human) 4856
Molecular Function:
growth factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support