Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: SAILGTEKST DGSKDVVEIA VPENLVGAIL GKGGKTLVEY QELTGARIQI SKKGEFVPGT RNRKVTITGT PAATQAAQYL ITQRITYEQG VRAANPQKVG
This gene encodes a neuron-specific RNA-binding protein, a member of the Nova family of paraneoplastic disease antigens, that is recognized and inhibited by paraneoplastic antibodies. These antibodies are found in the sera of patients with paraneoplastic opsoclonus-ataxia, breast cancer, and small cell lung cancer. Alternatively spliced transcripts encoding distinct isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuro-oncological ventral antigen 1; Onconeural ventral antigen 1; Paraneoplastic Ri antigen; RNA-binding protein Nova-1; Ventral neuron-specific protein 1
Gene Aliases: Nova-1; NOVA1
UniProt ID: (Human) P51513
Entrez Gene ID: (Human) 4857, (Rat) 298992
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support