Product References
Self-intensified synergy of a versatile biomimetic nanozyme and doxorubicin on electrospun fibers to inhibit postsurgical tumor recurrence and metastasis.
Biomaterials
Hu Y,Xu Y,Mintz RL,Luo X,Fang Y,Lao YH,Chan HF,Li K,Lv S,Chen G,Tao Y,Luo Y,Li M
Wed Feb 01 00:00:00 EST 2023
Self-intensified synergy of a versatile biomimetic nanozyme and doxorubicin on electrospun fibers to inhibit postsurgical tumor recurrence and metastasis.
Biomaterials
Hu Y,Xu Y,Mintz RL,Luo X,Fang Y,Lao YH,Chan HF,Li K,Lv S,Chen G,Tao Y,Luo Y,Li M
Wed Feb 01 00:00:00 EST 2023
iNOS contributes to heart failure with preserved ejection fraction through mitochondrial dysfunction and Akt S-nitrosylation.
Journal of advanced research
Guo Y,Wen J,He A,Qu C,Peng Y,Luo S,Wang X
Sun Jan 01 00:00:00 EST 2023
Exercise reduces hyperlipidemia-induced cardiac damage in apolipoprotein E-deficient mice via its effects against inflammation and oxidative stress.
Scientific reports
Pei Z,Ji J,Gao Y,Wang H,Wu Y,Yang J,Yang Q,Zhang L
Mon Jun 05 00:00:00 EDT 2023
Exercise reduces hyperlipidemia-induced cardiac damage in apolipoprotein E-deficient mice via its effects against inflammation and oxidative stress.
Scientific reports
Pei Z,Ji J,Gao Y,Wang H,Wu Y,Yang J,Yang Q,Zhang L
Mon Jun 05 00:00:00 EDT 2023
Blockade of Activin Receptor IIB Protects Arthritis Pathogenesis by Non-Amplification of Activin A-ACVR2B-NOX4 Axis Pathway.
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Jeon J,Lee H,Jeon MS,Kim SJ,Choi C,Kim KW,Yang DJ,Lee S,Bae YS,Choi WI,Jung J,Eyun SI,Yang S
Mon May 01 00:00:00 EDT 2023
Synergistic effects of combined treatment of 1,2-dichloroethane and high-dose ethanol on liver damage in mice and the related mechanisms.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Yang J,Zhang L,Wang T,Zhang J,Li M,Jin X,Tan X,Wang G,Zhao F,Jin Y
Thu Jun 01 00:00:00 EDT 2023
Synergistic effects of combined treatment of 1,2-dichloroethane and high-dose ethanol on liver damage in mice and the related mechanisms.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Yang J,Zhang L,Wang T,Zhang J,Li M,Jin X,Tan X,Wang G,Zhao F,Jin Y
Thu Jun 01 00:00:00 EDT 2023
Nodakenin Induces ROS-Dependent Apoptotic Cell Death and ER Stress in Radioresistant Breast Cancer.
Antioxidants (Basel, Switzerland)
Kim TW
Wed Feb 15 00:00:00 EST 2023
Elevated Angiotensin-II Levels Contribute to the Pathogenesis of Open-Angle Glaucoma Via Inducing the Expression of Fibrosis-Related Genes in Trabecular Meshwork Cells Through a ROS/NOX4/SMAD3 Axis.
Cell transplantation
Li H,Cui H,Ren J,Wang D,Zhao R,Zhu S,Liu S,Liu X,Tian S,Zhang Y,Zhao P,Li P,Thorne RF,Duan S
Mon Apr 03 00:00:00 EDT 2023
Nootkatone Supplementation Attenuates Carbon Tetrachloride Exposure-Induced Nephrotoxicity in Mice.
Antioxidants (Basel, Switzerland)
Dai C,Liu M,Zhang Q,Das Gupta S,Tang S,Shen J
Fri Feb 03 00:00:00 EST 2023
Irisin ameliorates D-galactose-induced skeletal muscle fibrosis via the PI3K/Akt pathway.
European journal of pharmacology
Wu Y,Wu Y,Yu J,Zhang Y,Li Y,Fu R,Sun Y,Zhao K,Xiao Q
Sun Jan 15 00:00:00 EST 2023
Acid sphingomyelinase promotes diabetic cardiomyopathy via NADPH oxidase 4 mediated apoptosis.
Cardiovascular diabetology
Liu R,Duan T,Yu L,Tang Y,Liu S,Wang C,Fang WJ
Thu Feb 02 00:00:00 EST 2023
Ursolic Acid Alleviates Cancer Cachexia and Prevents Muscle Wasting via Activating SIRT1.
Cancers
Tao W,Ouyang Z,Liao Z,Li L,Zhang Y,Gao J,Ma L,Yu S
Thu Apr 20 00:00:00 EDT 2023
Targeting ER Stress with Saikosaponin A to Overcome Resistance under Radiation in Gastric Cancer Cells.
International journal of molecular sciences
Kim TW
Thu Mar 16 00:00:00 EDT 2023
Synergistic effect of elevated glucose levels with SARS-CoV-2 spike protein induced NOX-dependent ROS production in endothelial cells.
Molecular biology reports
Zhang J,He L,Huang R,Alvarez JF,Yang DH,Sun Q,Wang F,Peng Z,Jiang N,Su L
Sat Jul 01 00:00:00 EDT 2023
Short-chain acyl-CoA dehydrogenase is a potential target for the treatment of vascular remodelling.
Journal of hypertension
Zhong X,Li Z,Xu Q,Peng H,Su Y,Le K,Shu Z,Liao Y,Ma Z,Pan X,Xu S,Zhou S
Mon May 01 00:00:00 EDT 2023
Adrenomedullin Improves Hypertension and Vascular Remodeling partly through the Receptor-Mediated AMPK Pathway in Rats with Obesity-Related Hypertension.
International journal of molecular sciences
Wang HY,Wang FZ,Chang R,Wang Q,Liu SY,Cheng ZX,Gao Q,Zhou H,Zhou YB
Wed Feb 15 00:00:00 EST 2023
Aloe-emodin ameliorated MI-induced cardiac remodeling in mice via inhibiting TGF-β/SMAD signaling via up-regulating SMAD7.
Phytomedicine : international journal of phytotherapy and phytopharmacology
Yu J,Zhao X,Yan X,Li W,Liu Y,Wang J,Wang J,Yang Y,Hao Y,Liang Z,Tao Y,Yuan Y,Du Z
Thu Jun 01 00:00:00 EDT 2023
Dexamethasone Induces Senescence-Associated Changes in Trabecular Meshwork Cells by Increasing ROS Levels Via the TGFβ/Smad3-NOX4 Axis.
Cell transplantation
Li H,Ren J,Cui H,Wang D,Zhao R,Liu X,Tian S,Wang J,Zhang J,Li P,Thorne RF,Duan S
Mon Jun 05 00:00:00 EDT 2023
Dexamethasone Induces Senescence-Associated Changes in Trabecular Meshwork Cells by Increasing ROS Levels Via the TGFβ/Smad3-NOX4 Axis.
Cell transplantation
Li H,Ren J,Cui H,Wang D,Zhao R,Liu X,Tian S,Wang J,Zhang J,Li P,Thorne RF,Duan S
Mon Jun 05 00:00:00 EDT 2023
Dexamethasone Induces Senescence-Associated Changes in Trabecular Meshwork Cells by Increasing ROS Levels Via the TGFβ/Smad3-NOX4 Axis.
Cell transplantation
Li H,Ren J,Cui H,Wang D,Zhao R,Liu X,Tian S,Wang J,Zhang J,Li P,Thorne RF,Duan S
Mon Jun 05 00:00:00 EDT 2023
Dexamethasone Induces Senescence-Associated Changes in Trabecular Meshwork Cells by Increasing ROS Levels Via the TGFβ/Smad3-NOX4 Axis.
Cell transplantation
Li H,Ren J,Cui H,Wang D,Zhao R,Liu X,Tian S,Wang J,Zhang J,Li P,Thorne RF,Duan S
Mon Jun 05 00:00:00 EDT 2023
Identifification and validation of ferroptosis signatures and immune infifiltration characteristics associated with intervertebral disc degeneration.
Frontiers in genetics
Zhang F,Cui D,Wang K,Cheng H,Zhai Y,Jiao W,Wang Z,Cui X,Yu H
Tue Mar 14 00:00:00 EDT 2023
Intermittent fasting alleviates type 1 diabetes-induced cognitive dysfunction by improving the frontal cortical metabolic disorder.
Biochimica et biophysica acta. Molecular basis of disease
Xiong F,Jiang K,Wu Y,Lou C,Ding C,Zhang W,Zhang X,Li C,Zheng H,Gao H
Sun Oct 01 00:00:00 EDT 2023
Effects of liraglutide on astrocyte polarization and neuroinflammation in db/db mice: focus on iron overload and oxidative stress.
Frontiers in cellular neuroscience
An JR,Liu JT,Gao XM,Wang QF,Sun GY,Su JN,Zhang C,Yu JX,Yang YF,Shi Y
Mon Jun 19 00:00:00 EDT 2023
Effects of apocynin on ISO-induced delayed afterdepolarizations in rat atrial myocytes and the underlying mechanisms.
Biochemical and biophysical research communications
Zhang N,Dan Q,Dong Y,Liu Y,Zhuang W,Xie Z,Zhao Q,Lin K,Li Y
Sun Jan 01 00:00:00 EST 2023
S-allylmercapto-N-acetylcysteine ameliorates pulmonary fibrosis in mice via Nrf2 pathway activation and NF-κB, TGF-β1/Smad2/3 pathway suppression.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhang Q,Ye W,Liu Y,Niu D,Zhao X,Li G,Qu Y,Zhao Z
Sun Jan 01 00:00:00 EST 2023
S-allylmercapto-N-acetylcysteine ameliorates pulmonary fibrosis in mice via Nrf2 pathway activation and NF-κB, TGF-β1/Smad2/3 pathway suppression.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhang Q,Ye W,Liu Y,Niu D,Zhao X,Li G,Qu Y,Zhao Z
Sun Jan 01 00:00:00 EST 2023
6-Shogaol Overcomes Gefitinib Resistance via ER Stress in Ovarian Cancer Cells.
International journal of molecular sciences
Kim TW,Lee HG
Mon Jan 30 00:00:00 EST 2023
Hypoxia-Induced Kidney Injury in Newborn Rats.
Toxics
Chu YT,Chen BH,Chen HH,Lee JC,Kuo TJ,Chiu HC,Lu WH
Sat Mar 11 00:00:00 EST 2023
NQO1 alleviates renal fibrosis by inhibiting the TLR4/NF-κB and TGF-β/Smad signaling pathways in diabetic nephropathy.
Cellular signalling
Qiu D,Song S,Chen N,Bian Y,Yuan C,Zhang W,Duan H,Shi Y
Tue Aug 01 00:00:00 EDT 2023
NQO1 alleviates renal fibrosis by inhibiting the TLR4/NF-κB and TGF-β/Smad signaling pathways in diabetic nephropathy.
Cellular signalling
Qiu D,Song S,Chen N,Bian Y,Yuan C,Zhang W,Duan H,Shi Y
Tue Aug 01 00:00:00 EDT 2023
Sirtuin 7 mitigates renal ferroptosis, fibrosis and injury in hypertensive mice by facilitating the KLF15/Nrf2 signaling.
Free radical biology & medicine
Li XT,Song JW,Zhang ZZ,Zhang MW,Liang LR,Miao R,Liu Y,Chen YH,Liu XY,Zhong JC
Sun Nov 20 00:00:00 EST 2022
Sirtuin 7 mitigates renal ferroptosis, fibrosis and injury in hypertensive mice by facilitating the KLF15/Nrf2 signaling.
Free radical biology & medicine
Li XT,Song JW,Zhang ZZ,Zhang MW,Liang LR,Miao R,Liu Y,Chen YH,Liu XY,Zhong JC
Sun Nov 20 00:00:00 EST 2022
The TBX1/miR-193a-3p/TGF-β2 Axis Mediates CHD by Promoting Ferroptosis.
Oxidative medicine and cellular longevity
Zhong L,Yang H,Zhu B,Zhao X,Xie M,Cao M,Liu C,Zhao D,Lyu Y,Shang W,Wang B,Wu Y,Sun X,Qiu G,Fu W,Jiang H
Fri Mar 04 00:00:00 EST 2022
MicroRNA 148a Suppresses Tuberculous Fibrosis by Targeting NOX4 and POLDIP2.
International journal of molecular sciences
Woo SJ,Kim Y,Jung H,Lee JJ,Hong JY
Thu Mar 10 00:00:00 EST 2022
Lonicera caerulea Berry Polyphenols Extract Alleviates Exercise Fatigue in Mice by Reducing Oxidative Stress, Inflammation, Skeletal Muscle Cell Apoptosis, and by Increasing Cell Proliferation.
Frontiers in nutrition
Liu S,Meng F,Zhang D,Shi D,Zhou J,Guo S,Chang X
Fri Apr 01 00:00:00 EDT 2022
Study on complications of osteoporosis based on network pharmacology.
Frontiers in genetics
Song Z,Zhang H,Jiang Y,Zhao R,Pei X,Ning H,Chen H,Pan J,Gong Y,Song M,Wang W
Wed Oct 19 00:00:00 EDT 2022
Rosmarinic Acid Ameliorates Pulmonary Ischemia/Reperfusion Injury by Activating the PI3K/Akt Signaling Pathway.
Frontiers in pharmacology
Luo W,Tao Y,Chen S,Luo H,Li X,Qu S,Chen K,Zeng C
Sat Jul 16 00:00:00 EDT 2022
Pyrrosia petiolosa Extract Ameliorates Ethylene Glycol-Induced Urolithiasis in Rats by Inhibiting Oxidative Stress and Inflammatory Response.
Disease markers
Zhou F,Wang X
Tue Aug 16 00:00:00 EDT 2022
Role of thioredoxin-interacting protein in mediating endothelial dysfunction in hypertension.
Genes & diseases
Wang R,Guo Y,Li L,Luo M,Peng L,Lv D,Cheng Z,Xue Q,Wang L,Huang J
Sun May 01 00:00:00 EDT 2022
Recurrent Hypoglycemia Impaired Vascular Function in Advanced T2DM Rats by Inducing Pyroptosis.
Oxidative medicine and cellular longevity
Luo M,Hu Y,Lv D,Xie L,Yang S,Zuo D,Xue Y,He A
Wed Aug 03 00:00:00 EDT 2022
HPV+ HNSCC-derived exosomal miR-9-5p inhibits TGF-β signaling-mediated fibroblast phenotypic transformation through NOX4.
Cancer science
Wang B,Zhang S,Tong F,Wang Y,Wei L
Fri Apr 01 00:00:00 EDT 2022
HPV+ HNSCC-derived exosomal miR-9-5p inhibits TGF-β signaling-mediated fibroblast phenotypic transformation through NOX4.
Cancer science
Wang B,Zhang S,Tong F,Wang Y,Wei L
Fri Apr 01 00:00:00 EDT 2022
The Relationship between Procyanidin Structure and Their Protective Effect in a Parkinson's Disease Model.
Molecules (Basel, Switzerland)
Chen J,Chen Y,Zheng Y,Zhao J,Yu H,Zhu J
Sat Aug 06 00:00:00 EDT 2022
Effect of Thymoquinone on Renal Damage Induced by Hyperlipidemia in LDL Receptor-Deficient (LDL-R-/-) Mice.
BioMed research international
Li W,Zhang H,Zhang L,Zhang T,Ding H
Tue Jul 19 00:00:00 EDT 2022
Effect of Thymoquinone on Renal Damage Induced by Hyperlipidemia in LDL Receptor-Deficient (LDL-R-/-) Mice.
BioMed research international
Li W,Zhang H,Zhang L,Zhang T,Ding H
Tue Jul 19 00:00:00 EDT 2022
Accumulation of advanced oxidation protein products contributes to age-related impairment of gap junction intercellular communication in osteocytes of male mice.
Bone & joint research
Tu C,Lai S,Huang Z,Cai G,Zhao K,Gao J,Wu Z,Zhong Z
Fri Jul 01 00:00:00 EDT 2022
Non-Invasive Local Acoustic Therapy Ameliorates Diabetic Heart Fibrosis by Suppressing ACE-Mediated Oxidative Stress and Inflammation in Cardiac Fibroblasts.
Cardiovascular drugs and therapy
Weng L,Li L,Zhao K,Xu T,Mao Y,Shu H,Chen X,Chen J,Wu J,Guo X,Tu J,Zhang D,Sun W,Kong X
Wed Jun 01 00:00:00 EDT 2022
Tumor necrosis factor-α coordinates with transforming growth factor-β1 to induce epithelial-mesenchymal transition and migration via the NF-κB/NOX4 pathway in bronchial epithelial cells.
Molecular biology reports
Li K,Zhou R,Ma M,Jin C,Jiao L,Zhang S,Tian M,Zhou F
Sat Oct 01 00:00:00 EDT 2022
Lysyl oxidase-like 2 inhibitor rescues D-galactose-induced skeletal muscle fibrosis.
Aging cell
Wu Y,Wu Y,Yang Y,Yu J,Wu J,Liao Z,Guo A,Sun Y,Zhao Y,Chen J,Xiao Q
Fri Jul 01 00:00:00 EDT 2022
5-Methoxyflavone alleviates LPS-mediated lung injury by promoting Nrf2-mediated the suppression of NOX4/TLR4 axis in bronchial epithelial cells and M1 polarization in macrophages.
Journal of inflammation (London, England)
Liang P,Wang L,Yang S,Pan X,Li J,Zhang Y,Liang Y,Li J,Zhou B
Wed Nov 30 00:00:00 EST 2022
5-Methoxyflavone alleviates LPS-mediated lung injury by promoting Nrf2-mediated the suppression of NOX4/TLR4 axis in bronchial epithelial cells and M1 polarization in macrophages.
Journal of inflammation (London, England)
Liang P,Wang L,Yang S,Pan X,Li J,Zhang Y,Liang Y,Li J,Zhou B
Wed Nov 30 00:00:00 EST 2022
5-Methoxyflavone alleviates LPS-mediated lung injury by promoting Nrf2-mediated the suppression of NOX4/TLR4 axis in bronchial epithelial cells and M1 polarization in macrophages.
Journal of inflammation (London, England)
Liang P,Wang L,Yang S,Pan X,Li J,Zhang Y,Liang Y,Li J,Zhou B
Wed Nov 30 00:00:00 EST 2022
5-Methoxyflavone alleviates LPS-mediated lung injury by promoting Nrf2-mediated the suppression of NOX4/TLR4 axis in bronchial epithelial cells and M1 polarization in macrophages.
Journal of inflammation (London, England)
Liang P,Wang L,Yang S,Pan X,Li J,Zhang Y,Liang Y,Li J,Zhou B
Wed Nov 30 00:00:00 EST 2022
Boosting cisplatin chemotherapy by nanomotor-enhanced tumor penetration and DNA adducts formation.
Journal of nanobiotechnology
Xu L,Zhang K,Ma X,Li Y,Jin Y,Liang C,Wang Y,Duan W,Zhang H,Zhang Z,Shi J,Liu J,Wang Y,Li W
Thu Sep 29 00:00:00 EDT 2022
Schisandrin B Diet Inhibits Oxidative Stress to Reduce Ferroptosis and Lipid Peroxidation to Prevent Pirarubicin-Induced Hepatotoxicity.
BioMed research international
Shi H,Yan Y,Yang H,Pu P,Tang H
Wed Sep 07 00:00:00 EDT 2022
Krill Oil Turns Off TGF-β1 Profibrotic Signaling in the Prevention of Diabetic Nephropathy.
Journal of agricultural and food chemistry
Sun X,Yang Y,Sun X,Meng H,Hao W,Yin J,Ma F,Guo X,Du L,Sun L,Wu H
Wed Aug 17 00:00:00 EDT 2022
PRMT5 critically mediates TMAO-induced inflammatory response in vascular smooth muscle cells.
Cell death & disease
Liu H,Jia K,Ren Z,Sun J,Pan LL
Mon Apr 04 00:00:00 EDT 2022
Melatonin Attenuates Dasatinib-Aggravated Hypoxic Pulmonary Hypertension via Inhibiting Pulmonary Vascular Remodeling.
Frontiers in cardiovascular medicine
Wang R,Pan J,Han J,Gong M,Liu L,Zhang Y,Liu Y,Wang D,Tang Q,Wu N,Wang L,Yan J,Li H,Yuan Y
Wed Apr 13 00:00:00 EDT 2022
Aerobic Exercise Training Improves Renal Injury in Spontaneously Hypertensive Rats by Increasing Renalase Expression in Medulla.
Frontiers in cardiovascular medicine
Luo M,Cao S,Lv D,He L,He Z,Li L,Li Y,Luo S,Chang Q
Fri Jul 29 00:00:00 EDT 2022
Pu-erh Tea Restored Circadian Rhythm Disruption by Regulating Tryptophan Metabolism.
Journal of agricultural and food chemistry
Hu S,Luo L,Bian X,Liu RH,Zhao S,Chen Y,Sun K,Jiang J,Liu Z,Zeng L
Wed May 11 00:00:00 EDT 2022
Hypobaric hypoxia triggers pyroptosis in the retina via NLRP3 inflammasome activation.
Apoptosis : an international journal on programmed cell death
Xin X,Yang K,Liu H,Li Y
Fri Apr 01 00:00:00 EDT 2022
Hypobaric hypoxia triggers pyroptosis in the retina via NLRP3 inflammasome activation.
Apoptosis : an international journal on programmed cell death
Xin X,Yang K,Liu H,Li Y
Fri Apr 01 00:00:00 EDT 2022
Bromelain Ameliorates Atherosclerosis by Activating the TFEB-Mediated Autophagy and Antioxidant Pathways.
Antioxidants (Basel, Switzerland)
Chen CH,Hsia CC,Hu PA,Yeh CH,Chen CT,Peng CL,Wang CH,Lee TS
Thu Dec 29 00:00:00 EST 2022
NOX4 Mediates Epithelial Cell Death in Hyperoxic Acute Lung Injury Through Mitochondrial Reactive Oxygen Species.
Frontiers in pharmacology
Harijith A,Basa P,Ha A,Thomas J,Jafri A,Fu P,MacFarlane PM,Raffay TM,Natarajan V,Sudhadevi T
Sat Jul 16 00:00:00 EDT 2022
NOX4 Mediates Epithelial Cell Death in Hyperoxic Acute Lung Injury Through Mitochondrial Reactive Oxygen Species.
Frontiers in pharmacology
Harijith A,Basa P,Ha A,Thomas J,Jafri A,Fu P,MacFarlane PM,Raffay TM,Natarajan V,Sudhadevi T
Sat Jul 16 00:00:00 EDT 2022
Effects of thymoquinone against angiotensin II‑induced cardiac damage in apolipoprotein E‑deficient mice.
International journal of molecular medicine
Zhang L,Zhang H,Ma J,Wang Y,Pei Z,Ding H
Sun May 01 00:00:00 EDT 2022
Effects of thymoquinone against angiotensin II‑induced cardiac damage in apolipoprotein E‑deficient mice.
International journal of molecular medicine
Zhang L,Zhang H,Ma J,Wang Y,Pei Z,Ding H
Sun May 01 00:00:00 EDT 2022
Effects of thymoquinone against angiotensin II‑induced cardiac damage in apolipoprotein E‑deficient mice.
International journal of molecular medicine
Zhang L,Zhang H,Ma J,Wang Y,Pei Z,Ding H
Sun May 01 00:00:00 EDT 2022
Effects of thymoquinone against angiotensin II‑induced cardiac damage in apolipoprotein E‑deficient mice.
International journal of molecular medicine
Zhang L,Zhang H,Ma J,Wang Y,Pei Z,Ding H
Sun May 01 00:00:00 EDT 2022
Jujuboside A ameliorates high fat diet and streptozotocin induced diabetic nephropathy via suppressing oxidative stress, apoptosis, and enhancing autophagy.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Zhong Y,Luo R,Liu Q,Zhu J,Lei M,Liang X,Wang X,Peng X
Sat Jan 01 00:00:00 EST 2022
Suramin enhances chondrogenic properties by regulating the p67phox/PI3K/AKT/SOX9 signalling pathway.
Bone & joint research
Liu ZM,Shen PC,Lu CC,Chou SH,Tien YC
Sat Oct 01 00:00:00 EDT 2022
Sirtuin1 inhibitor attenuates hypertension in spontaneously hypertensive rats: role of Giα proteins and nitroxidative stress.
Journal of hypertension
Arifen N,Li Y,Srivastava AK,Anand-Srivastava MB
Fri Jul 01 00:00:00 EDT 2022
Impact of Selective Renal Afferent Denervation on Oxidative Stress and Vascular Remodeling in Spontaneously Hypertensive Rats.
Antioxidants (Basel, Switzerland)
Wu LL,Zhang Y,Li XZ,Du XL,Gao Y,Wang JX,Wang XL,Chen Q,Li YH,Zhu GQ,Tan X
Fri May 20 00:00:00 EDT 2022
Angiotensin II Induces Cardiac Edema and Hypertrophic Remodeling through Lymphatic-Dependent Mechanisms.
Oxidative medicine and cellular longevity
Bai J,Yin L,Yu WJ,Zhang YL,Lin QY,Li HH
Thu Mar 17 00:00:00 EDT 2022
Astragaloside IV ameliorates sepsis-induced myocardial dysfunction by regulating NOX4/JNK/BAX pathway.
Life sciences
Su Y,Yin X,Huang X,Guo Q,Ma M,Guo L
Thu Dec 01 00:00:00 EST 2022
SIRT7 Deficiency Protects against Aβ42-Induced Apoptosis through the Regulation of NOX4-Derived Reactive Oxygen Species Production in SH-SY5Y Cells.
International journal of molecular sciences
Mizutani H,Sato Y,Yamazaki M,Yoshizawa T,Ando Y,Ueda M,Yamagata K
Fri Aug 12 00:00:00 EDT 2022
In utero hypoxia attenuated acetylcholine-mediated vasodilatation via CHRM3/p-NOS3 in fetal sheep MCA: role of ROS/ERK1/2.
Hypertension research : official journal of the Japanese Society of Hypertension
He Y,Su H,Li N,Zhang Y,Zhang P,Zhang Y,Ye Y,Zhang Y,Tang J,Xu Z
Fri Jul 01 00:00:00 EDT 2022
Effect of thymoquinone on sepsis-induced cardiac damage via anti-inflammatory and anti-apoptotic mechanisms.
The Journal of international medical research
Guo W,Long X,Lv M,Deng S,Liu D,Yang Q
Thu Sep 01 00:00:00 EDT 2022
Comprehensive analysis of placental gene-expression profiles and identification of EGFR-mediated autophagy and ferroptosis suppression in intrahepatic cholestasis of pregnancy.
Gene
Fang Y,Fang D
Fri Aug 05 00:00:00 EDT 2022
Intimate intertwining of the pathogenesis of hypoxia and systemic sclerosis: A transcriptome integration analysis.
Frontiers in immunology
He X,Shi Y,Zeng Z,Tang B,Xiao X,Yu J,Zou P,Liu J,Xiao Y,Luo Y,Xiao R
Mon Nov 21 00:00:00 EST 2022
Sphingosine-1-Phosphate Alleviates Irradiation Induced Salivary Gland Hypofunction through Preserving Endothelial Cells and Resident Macrophages.
Antioxidants (Basel, Switzerland)
Yang T,Zhao Q,Hu M,Pan S,Zhang L,Zhu R,Zhou B,Feng X,Gao Z,Zhu Z,Zhang Y,Hu L,Liu F,Shan Z
Tue Oct 18 00:00:00 EDT 2022
Colistin-induced pulmonary toxicity involves the activation of NOX4/TGF-β/mtROS pathway and the inhibition of Akt/mTOR pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Dai C,Li M,Sun T,Zhang Y,Wang Y,Shen Z,Velkov T,Tang S,Shen J
Sun May 01 00:00:00 EDT 2022
Colistin-induced pulmonary toxicity involves the activation of NOX4/TGF-β/mtROS pathway and the inhibition of Akt/mTOR pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Dai C,Li M,Sun T,Zhang Y,Wang Y,Shen Z,Velkov T,Tang S,Shen J
Sun May 01 00:00:00 EDT 2022
Colistin-induced pulmonary toxicity involves the activation of NOX4/TGF-β/mtROS pathway and the inhibition of Akt/mTOR pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Dai C,Li M,Sun T,Zhang Y,Wang Y,Shen Z,Velkov T,Tang S,Shen J
Sun May 01 00:00:00 EDT 2022
Colistin-induced pulmonary toxicity involves the activation of NOX4/TGF-β/mtROS pathway and the inhibition of Akt/mTOR pathway.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Dai C,Li M,Sun T,Zhang Y,Wang Y,Shen Z,Velkov T,Tang S,Shen J
Sun May 01 00:00:00 EDT 2022
A Novel Compound, Tanshinol Borneol Ester, Ameliorates Pressure Overload-Induced Cardiac Hypertrophy by Inhibiting Oxidative Stress via the mTOR/β-TrCP/NRF2 Pathway.
Frontiers in pharmacology
Han D,Wang F,Wang B,Qiao Z,Cui X,Zhang Y,Jiang Q,Liu M,Shangguan J,Zheng X,Bai Y,Du C,Shen D
Wed Feb 23 00:00:00 EST 2022
NAD(P)H: quinone oxidoreductase 1 attenuates oxidative stress and apoptosis by regulating Sirt1 in diabetic nephropathy.
Journal of translational medicine
Qiu D,Song S,Wang Y,Bian Y,Wu M,Wu H,Shi Y,Duan H
Fri Jan 28 00:00:00 EST 2022
NAD(P)H: quinone oxidoreductase 1 attenuates oxidative stress and apoptosis by regulating Sirt1 in diabetic nephropathy.
Journal of translational medicine
Qiu D,Song S,Wang Y,Bian Y,Wu M,Wu H,Shi Y,Duan H
Fri Jan 28 00:00:00 EST 2022
MicroRNA-122-5p promotes renal fibrosis and injury in spontaneously hypertensive rats by targeting FOXO3.
Experimental cell research
Liu Y,Dong ZJ,Song JW,Liang LR,Sun LL,Liu XY,Miao R,Xu YL,Li XT,Zhang MW,Zhang ZZ,Zhong JC
Tue Feb 15 00:00:00 EST 2022
MicroRNA-122-5p promotes renal fibrosis and injury in spontaneously hypertensive rats by targeting FOXO3.
Experimental cell research
Liu Y,Dong ZJ,Song JW,Liang LR,Sun LL,Liu XY,Miao R,Xu YL,Li XT,Zhang MW,Zhang ZZ,Zhong JC
Tue Feb 15 00:00:00 EST 2022
MicroRNA-122-5p promotes renal fibrosis and injury in spontaneously hypertensive rats by targeting FOXO3.
Experimental cell research
Liu Y,Dong ZJ,Song JW,Liang LR,Sun LL,Liu XY,Miao R,Xu YL,Li XT,Zhang MW,Zhang ZZ,Zhong JC
Tue Feb 15 00:00:00 EST 2022
MicroRNA-122-5p promotes renal fibrosis and injury in spontaneously hypertensive rats by targeting FOXO3.
Experimental cell research
Liu Y,Dong ZJ,Song JW,Liang LR,Sun LL,Liu XY,Miao R,Xu YL,Li XT,Zhang MW,Zhang ZZ,Zhong JC
Tue Feb 15 00:00:00 EST 2022
Sestrin2 remedies podocyte injury via orchestrating TSP-1/TGF-β1/Smad3 axis in diabetic kidney disease.
Cell death & disease
Song S,Shi C,Bian Y,Yang Z,Mu L,Wu H,Duan H,Shi Y
Sat Jul 30 00:00:00 EDT 2022
Connexin32 ameliorates epithelial-to-mesenchymal-transition in diabetic renal tubular via inhibiting NOX4.
Pharmacological research
Sun X,Xiao H,Li S,Chen R,Lin Z,Yang Y,Chen Z,Deng L,Huang H
Tue Feb 01 00:00:00 EST 2022
Connexin32 ameliorates epithelial-to-mesenchymal-transition in diabetic renal tubular via inhibiting NOX4.
Pharmacological research
Sun X,Xiao H,Li S,Chen R,Lin Z,Yang Y,Chen Z,Deng L,Huang H
Tue Feb 01 00:00:00 EST 2022
Connexin32 ameliorates epithelial-to-mesenchymal-transition in diabetic renal tubular via inhibiting NOX4.
Pharmacological research
Sun X,Xiao H,Li S,Chen R,Lin Z,Yang Y,Chen Z,Deng L,Huang H
Tue Feb 01 00:00:00 EST 2022
Connexin32 ameliorates epithelial-to-mesenchymal-transition in diabetic renal tubular via inhibiting NOX4.
Pharmacological research
Sun X,Xiao H,Li S,Chen R,Lin Z,Yang Y,Chen Z,Deng L,Huang H
Tue Feb 01 00:00:00 EST 2022
Connexin32 ameliorates epithelial-to-mesenchymal-transition in diabetic renal tubular via inhibiting NOX4.
Pharmacological research
Sun X,Xiao H,Li S,Chen R,Lin Z,Yang Y,Chen Z,Deng L,Huang H
Tue Feb 01 00:00:00 EST 2022
Connexin32 ameliorates epithelial-to-mesenchymal-transition in diabetic renal tubular via inhibiting NOX4.
Pharmacological research
Sun X,Xiao H,Li S,Chen R,Lin Z,Yang Y,Chen Z,Deng L,Huang H
Tue Feb 01 00:00:00 EST 2022
Lpcat3 deficiency promotes palmitic acid-induced 3T3-L1 mature adipocyte inflammation through enhanced ROS generation.
Acta biochimica et biophysica Sinica
Hu J,Deng Y,Ding T,Dong J,Liang Y,Lou B
Fri Nov 25 00:00:00 EST 2022
Fluorofenidone Inhibits UUO/IRI-Induced Renal Fibrosis by Reducing Mitochondrial Damage.
Oxidative medicine and cellular longevity
Liao X,Lv X,Zhang Y,Han Y,Li J,Zeng J,Tang D,Meng J,Yuan X,Peng Z,Tao L,Xie Y
Fri Apr 01 00:00:00 EDT 2022
Sestrin2 attenuates renal damage by regulating Hippo pathway in diabetic nephropathy.
Cell and tissue research
Bian Y,Shi C,Song S,Mu L,Wu M,Qiu D,Dong J,Zhang W,Yuan C,Wang D,Zhou Z,Dong X,Shi Y
Sat Oct 01 00:00:00 EDT 2022
Sestrin2 attenuates renal damage by regulating Hippo pathway in diabetic nephropathy.
Cell and tissue research
Bian Y,Shi C,Song S,Mu L,Wu M,Qiu D,Dong J,Zhang W,Yuan C,Wang D,Zhou Z,Dong X,Shi Y
Sat Oct 01 00:00:00 EDT 2022
The BCL2/BAX/ROS pathway is involved in the inhibitory effect of astragaloside IV on pyroptosis in human umbilical vein endothelial cells.
Pharmaceutical biology
Su Y,Yin X,Huang X,Guo Q,Ma M,Guo L
Thu Dec 01 00:00:00 EST 2022
Ferroptosis-related gene NOX4, CHAC1 and HIF1A are valid biomarkers for stomach adenocarcinoma.
Journal of cellular and molecular medicine
Xiao R,Wang S,Guo J,Liu S,Ding A,Wang G,Li W,Zhang Y,Bian X,Zhao S,Qiu W
Tue Feb 01 00:00:00 EST 2022
Apelin-13 Attenuates Lipopolysaccharide-Induced Inflammatory Responses and Acute Lung Injury by Regulating PFKFB3-Driven Glycolysis Induced by NOX4-Dependent ROS.
Journal of inflammation research
Yuan Y,Wang W,Zhang Y,Hong Q,Huang W,Li L,Xie Z,Chen Y,Li X,Meng Y
Fri Apr 08 00:00:00 EDT 2022
Apelin-13 Attenuates Lipopolysaccharide-Induced Inflammatory Responses and Acute Lung Injury by Regulating PFKFB3-Driven Glycolysis Induced by NOX4-Dependent ROS.
Journal of inflammation research
Yuan Y,Wang W,Zhang Y,Hong Q,Huang W,Li L,Xie Z,Chen Y,Li X,Meng Y
Fri Apr 08 00:00:00 EDT 2022
Radioprotective effects and mechanism of HL-003 on radiation-induced salivary gland damage in mice.
Scientific reports
Ren J,Huang R,Li Y,Chen R,Tian H,Liu C
Thu May 19 00:00:00 EDT 2022
Ageing Increases Cardiac Electrical Remodelling in Rats and Mice via NOX4/ROS/CaMKII-Mediated Calcium Signalling.
Oxidative medicine and cellular longevity
Luo X,Yu W,Liu Z,Pu Z,Liu T,Li Y,Liu W,Lei M,Tan X,Chen T
Fri Apr 08 00:00:00 EDT 2022
Effects of β-glucan, probiotics, and synbiotics on obesity-associated colitis and hepatic manifestations in C57BL/6J mice.
European journal of nutrition
Vu V,Muthuramalingam K,Singh V,Hyun C,Kim YM,Unno T,Cho M
Tue Mar 01 00:00:00 EST 2022
Blocking REDD1/TXNIP Complex Ameliorates HG-Induced Renal Tubular Epithelial Cell Apoptosis and EMT through Repressing Oxidative Stress.
International journal of endocrinology
Mu L,Chen N,Chen Y,Yang Z,Zhou H,Song S,Shi Y
Tue Oct 04 00:00:00 EDT 2022
MST1/2 inhibitor XMU-MP-1 alleviates the injury induced by ionizing radiation in haematopoietic and intestinal system.
Journal of cellular and molecular medicine
Zhou X,Wang H,Li D,Song N,Yang F,Xu W
Tue Mar 01 00:00:00 EST 2022
ZFP36 protects against oxygen-glucose deprivation/reoxygenation-induced mitochondrial fragmentation and neuronal apoptosis through inhibiting NOX4-DRP1 pathway.
Brain research bulletin
Guo H,Jiang Y,Gu Z,Ren L,Zhu C,Yu S,Wei R
Tue Feb 01 00:00:00 EST 2022
Dioscin relieves diabetic nephropathy via suppressing oxidative stress and apoptosis, and improving mitochondrial quality and quantity control.
Food & function
Zhong Y,Liu J,Sun D,Guo T,Yao Y,Xia X,Shi C,Peng X
Mon Mar 21 00:00:00 EDT 2022
Administration of USP7 inhibitor P22077 inhibited cardiac hypertrophy and remodeling in Ang II-induced hypertensive mice.
Frontiers in pharmacology
Gu YH,Ren KW,Wang Y,Wang SH,Yu XH,Xu LW,Li HH,Bi HL
Fri Nov 18 00:00:00 EST 2022
Soluble Epoxide Hydrolase Inhibition Protected against Diabetic Cardiomyopathy through Inducing Autophagy and Reducing Apoptosis Relying on Nrf2 Upregulation and Transcription Activation.
Oxidative medicine and cellular longevity
Fang Q,Liu X,Ding J,Zhang Z,Chen G,Du T,Wang Y,Xu R
Wed Apr 06 00:00:00 EDT 2022
Human Tissue Kallikrein 1 Is Downregulated in Elderly Human Prostates and Possesses Potential In Vitro Antioxidative and Antifibrotic Effects in Rodent Prostates.
Oxidative medicine and cellular longevity
Zhang M,Luo C,Lin D,Cui K,Chen Z,Liu J
Fri Nov 12 00:00:00 EST 2021
Activated Factor X Signaling Pathway via Protease-Activated Receptor 2 Is a Novel Therapeutic Target for Preventing Atrial Fibrillation.
Circulation journal : official journal of the Japanese Circulation Society
Matsuura T,Soeki T,Fukuda D,Uematsu E,Tobiume T,Hara T,Kusunose K,Ise T,Yamaguchi K,Yagi S,Yamada H,Wakatsuki T,Sata M
Wed Jul 21 00:00:00 EDT 2021
17β-Estradiol Inhibits Proliferation and Oxidative Stress in Vascular Smooth Muscle Cells by Upregulating BHLHE40 Expression.
Frontiers in cardiovascular medicine
Feng DD,Zheng B,Yu J,Zhang ML,Ma Y,Hao X,Wen JK,Zhang XH
Sat Dec 18 00:00:00 EST 2021
Melatonin Alleviates Contrast-Induced Acute Kidney Injury by Activation of Sirt3.
Oxidative medicine and cellular longevity
Zhang C,Suo M,Liu L,Qi Y,Zhang C,Xie L,Zheng X,Ma C,Li J,Yang J,Bu P
Wed Dec 29 00:00:00 EST 2021
ChREBP deficiency alleviates apoptosis by inhibiting TXNIP/oxidative stress in diabetic nephropathy.
Journal of diabetes and its complications
Chen N,Song S,Yang Z,Wu M,Mu L,Zhou T,Shi Y
Wed Dec 01 00:00:00 EST 2021
ChREBP deficiency alleviates apoptosis by inhibiting TXNIP/oxidative stress in diabetic nephropathy.
Journal of diabetes and its complications
Chen N,Song S,Yang Z,Wu M,Mu L,Zhou T,Shi Y
Wed Dec 01 00:00:00 EST 2021
ChREBP deficiency alleviates apoptosis by inhibiting TXNIP/oxidative stress in diabetic nephropathy.
Journal of diabetes and its complications
Chen N,Song S,Yang Z,Wu M,Mu L,Zhou T,Shi Y
Wed Dec 01 00:00:00 EST 2021
ChREBP deficiency alleviates apoptosis by inhibiting TXNIP/oxidative stress in diabetic nephropathy.
Journal of diabetes and its complications
Chen N,Song S,Yang Z,Wu M,Mu L,Zhou T,Shi Y
Wed Dec 01 00:00:00 EST 2021
Sirtuin 1 ameliorates defenestration in hepatic sinusoidal endothelial cells during liver fibrosis via inhibiting stress-induced premature senescence.
Cell proliferation
Luo X,Bai Y,He S,Sun S,Jiang X,Yang Z,Lu D,Wei P,Liang Y,Peng C,Wang Y,Sheng R,Han S,Li X,Zhang B
Mon Mar 01 00:00:00 EST 2021
Osthole Attenuates Bleomycin-Induced Pulmonary Fibrosis by Modulating NADPH Oxidase 4-Derived Oxidative Stress in Mice.
Oxidative medicine and cellular longevity
Fang L,Wang W,Chen J,Zuo A,Gao H,Yan T,Wang P,Lu Y,Lv R,Xu F,Chen Y,Lyu L
Thu Jan 20 00:00:00 EST 2022
Blue LED causes autophagic cell death in human osteosarcoma by increasing ROS generation and dephosphorylating EGFR.
Journal of cellular and molecular medicine
He M,Yan G,Wang Y,Gong R,Lei H,Yu S,He X,Li G,Du W,Ma T,Gao M,Yu M,Liu S,Xu Z,Idiiatullina E,Zagidullin N,Pavlov V,Cai B,Yuan Y,Yang L
Tue Jun 01 00:00:00 EDT 2021
RND3 attenuates oxidative stress and vascular remodeling in spontaneously hypertensive rat via inhibiting ROCK1 signaling.
Redox biology
Wu N,Zheng F,Li N,Han Y,Xiong XQ,Wang JJ,Chen Q,Li YH,Zhu GQ,Zhou YB
Mon Dec 06 00:00:00 EST 2021
Silencing COX-2 blocks PDK1/TRAF4-induced AKT activation to inhibit fibrogenesis during skeletal muscle atrophy.
Redox biology
Chen H,Qian Z,Zhang S,Tang J,Fang L,Jiang F,Ge D,Chang J,Cao J,Yang L,Cao X
Fri Jan 01 00:00:00 EST 2021
Silencing COX-2 blocks PDK1/TRAF4-induced AKT activation to inhibit fibrogenesis during skeletal muscle atrophy.
Redox biology
Chen H,Qian Z,Zhang S,Tang J,Fang L,Jiang F,Ge D,Chang J,Cao J,Yang L,Cao X
Fri Jan 01 00:00:00 EST 2021
A neutral polysaccharide from Ophiocordyceps lanpingensis restrains cisplatin-induced nephrotoxicity.
Food science & nutrition
Zhou S,Zhou Y,Yu J,Jiang L,Xiang Y,Wang J,Du Y,Cui X,Ge F
Thu Jul 01 00:00:00 EDT 2021
Oxidative Stress-Induced Sirtuin1 Downregulation Correlates to HIF-1α, GLUT-1, and VEGF-A Upregulation in Th1 Autoimmune Hashimoto's Thyroiditis.
International journal of molecular sciences
Hepp M,Werion A,De Greef A,de Ville de Goyet C,de Bournonville M,Behets C,Lengelé B,Daumerie C,Mourad M,Ludgate M,Many MC,Joris V,Craps J
Wed Apr 07 00:00:00 EDT 2021
Oxidative Stress-Induced Sirtuin1 Downregulation Correlates to HIF-1α, GLUT-1, and VEGF-A Upregulation in Th1 Autoimmune Hashimoto's Thyroiditis.
International journal of molecular sciences
Hepp M,Werion A,De Greef A,de Ville de Goyet C,de Bournonville M,Behets C,Lengelé B,Daumerie C,Mourad M,Ludgate M,Many MC,Joris V,Craps J
Wed Apr 07 00:00:00 EDT 2021
Effects of different intensities of continuous training on vascular inflammation and oxidative stress in spontaneously hypertensive rats.
Journal of cellular and molecular medicine
Luo M,Cao C,Niebauer J,Yan J,Ma X,Chang Q,Zhang T,Huang X,Liu G
Wed Sep 01 00:00:00 EDT 2021
Exercise Intervention Mitigates Pathological Liver Changes in NAFLD Zebrafish by Activating SIRT1/AMPK/NRF2 Signaling.
International journal of molecular sciences
Zou Y,Chen Z,Sun C,Yang D,Zhou Z,Peng X,Zheng L,Tang C
Sun Oct 10 00:00:00 EDT 2021
The effects of compound centella formula on OxInflammation and silent information regulator 1 in a high-fat diet/streptozotocin-induced diabetic kidney disease rat model.
Experimental and therapeutic medicine
Zhu Q,Li XH,Chen HY,Jin QY
Wed Sep 01 00:00:00 EDT 2021
LRRC8A contributes to angiotensin II-induced cardiac hypertrophy by interacting with NADPH oxidases via the C-terminal leucine-rich repeat domain.
Free radical biology & medicine
Huo C,Liu Y,Li X,Xu R,Jia X,Hou L,Wang X
Mon Mar 01 00:00:00 EST 2021
IRE1α/NOX4 signaling pathway mediates ROS-dependent activation of hepatic stellate cells in NaAsO2 -induced liver fibrosis.
Journal of cellular physiology
Tao Y,Qiu T,Yao X,Jiang L,Wang N,Jiang J,Jia X,Wei S,Zhang J,Zhu Y,Tian W,Yang G,Liu X,Liu S,Ding Y,Sun X
Mon Feb 01 00:00:00 EST 2021
TGF-β promote epithelial-mesenchymal transition via NF-κB/NOX4/ROS signal pathway in lung cancer cells.
Molecular biology reports
Ma M,Shi F,Zhai R,Wang H,Li K,Xu C,Yao W,Zhou F
Mon Mar 01 00:00:00 EST 2021
Macrophagic Extracellular Vesicle CXCL2 Recruits and Activates the Neutrophil CXCR2/PKC/NOX4 Axis in Sepsis.
Journal of immunology (Baltimore, Md. : 1950)
Wang G,Huang W,Wang S,Wang J,Cui W,Zhang W,Lou A,Geng S,Li X
Fri Oct 15 00:00:00 EDT 2021
miR-199a Downregulation as a Driver of the NOX4/HIF-1α/VEGF-A Pathway in Thyroid and Orbital Adipose Tissues from Graves' Patients.
International journal of molecular sciences
Craps J,Joris V,Baldeschi L,Daumerie C,Camboni A,Buemi A,Lengelé B,Behets C,Boschi A,Mourad M,Many MC,Dessy C
Thu Dec 23 00:00:00 EST 2021
miR-199a Downregulation as a Driver of the NOX4/HIF-1α/VEGF-A Pathway in Thyroid and Orbital Adipose Tissues from Graves' Patients.
International journal of molecular sciences
Craps J,Joris V,Baldeschi L,Daumerie C,Camboni A,Buemi A,Lengelé B,Behets C,Boschi A,Mourad M,Many MC,Dessy C
Thu Dec 23 00:00:00 EST 2021
Amelioration of Juglanin against LPS-Induced Activation of NLRP3 Inflammasome in Chondrocytes Mediated by SIRT1.
Inflammation
Wang T,Wang J,Sun T,Li Y
Tue Jun 01 00:00:00 EDT 2021
Aged Ripe Pu-erh Tea Reduced Oxidative Stress-Mediated Inflammation in Dextran Sulfate Sodium-Induced Colitis Mice by Regulating Intestinal Microbes.
Journal of agricultural and food chemistry
Hu S,Li S,Liu Y,Sun K,Luo L,Zeng L
Wed Sep 15 00:00:00 EDT 2021
miR-31-5p Promotes Oxidative Stress and Vascular Smooth Muscle Cell Migration in Spontaneously Hypertensive Rats via Inhibiting FNDC5 Expression.
Biomedicines
Zhou B,Wu LL,Zheng F,Wu N,Chen AD,Zhou H,Chen JY,Chen Q,Li YH,Kang YM,Zhu GQ
Fri Aug 13 00:00:00 EDT 2021
The protective effect of harpagoside on angiotensin II (Ang II)-induced blood-brain barrier leakage in vitro.
Phytotherapy research : PTR
Lu YW,Hao RJ,Wei YY,Yu GR
Mon Nov 01 00:00:00 EDT 2021
Diabetes Promotes Retinal Vascular Endothelial Cell Injury by Inducing CCN1 Expression.
Frontiers in cardiovascular medicine
Li H,Li T,Wang H,He X,Li Y,Wen S,Peng R,Nie Y,Lu Y,Yang H,Ye Y,Shi G,Chen Y
Tue Aug 31 00:00:00 EDT 2021
Diabetes Promotes Retinal Vascular Endothelial Cell Injury by Inducing CCN1 Expression.
Frontiers in cardiovascular medicine
Li H,Li T,Wang H,He X,Li Y,Wen S,Peng R,Nie Y,Lu Y,Yang H,Ye Y,Shi G,Chen Y
Tue Aug 31 00:00:00 EDT 2021
C-C motif ligand 8 promotes atherosclerosis via NADPH oxidase 2/reactive oxygen species-induced endothelial permeability increase.
Free radical biology & medicine
Xue S,Tang H,Zhao G,Fang C,Shen Y,Yan D,Yuan Y,Fu W,Shi Z,Tang X,Guo D
Sat May 01 00:00:00 EDT 2021
Inhibition of NLRP3 inflammasome ameliorates podocyte damage by suppressing lipid accumulation in diabetic nephropathy.
Metabolism: clinical and experimental
Wu M,Yang Z,Zhang C,Shi Y,Han W,Song S,Mu L,Du C,Shi Y
Sat May 01 00:00:00 EDT 2021
Carbohydrate response element-binding protein regulates lipid metabolism via mTOR complex1 in diabetic nephropathy.
Journal of cellular physiology
Chen N,Mu L,Yang Z,Du C,Wu M,Song S,Yuan C,Shi Y
Fri Jan 01 00:00:00 EST 2021
Artemisinin Improves Acetylcholine-Induced Vasodilatation in Rats with Primary Hypertension.
Drug design, development and therapy
Liu X,Wang X,Pan Y,Zhao L,Sun S,Luo A,Bao C,Tang H,Han Y
Tue Feb 22 00:00:00 EST 2022
The Herbal Formula JI017 Induces ER Stress via Nox4 in Breast Cancer Cells.
Antioxidants (Basel, Switzerland)
Kim TW,Ko SG
Thu Nov 25 00:00:00 EST 2021
Extracellular Vesicles Derived From Human Umbilical Cord Mesenchymal Stem Cells Protect Against DOX-Induced Heart Failure Through the miR-100-5p/NOX4 Pathway.
Frontiers in bioengineering and biotechnology
Zhong Z,Tian Y,Luo X,Zou J,Wu L,Tian J
Tue Sep 14 00:00:00 EDT 2021
NOX4 Mediates Pseudomonas aeruginosa-Induced Nuclear Reactive Oxygen Species Generation and Chromatin Remodeling in Lung Epithelium.
Antioxidants (Basel, Switzerland)
Fu P,Ramchandran R,Sudhadevi T,Kumar PPK,Krishnan Y,Liu Y,Zhao Y,Parinandi NL,Harijith A,Sadoshima J,Natarajan V
Wed Mar 17 00:00:00 EDT 2021
Scoparone alleviates Ang II-induced pathological myocardial hypertrophy in mice by inhibiting oxidative stress.
Journal of cellular and molecular medicine
Lyu L,Chen J,Wang W,Yan T,Lin J,Gao H,Li H,Lv R,Xu F,Fang L,Chen Y
Mon Mar 01 00:00:00 EST 2021
Lack of Endothelial Nitric Oxide Synthase Accelerates Ectopic Calcification in Uremic Mice Fed an Adenine and High Phosphorus Diet.
The American journal of pathology
Oe Y,Mitsui S,Sato E,Shibata N,Kisu K,Sekimoto A,Miyazaki M,Sato H,Ito S,Takahashi N
Mon Feb 01 00:00:00 EST 2021
Melatonin Alleviates Renal Injury in Mouse Model of Sepsis.
Frontiers in pharmacology
Chen L,Han Z,Shi Z,Liu C,Lu Q
Tue Sep 21 00:00:00 EDT 2021
Nox4 Promotes RANKL-Induced Autophagy and Osteoclastogenesis via Activating ROS/PERK/eIF-2α/ATF4 Pathway.
Frontiers in pharmacology
Sun J,Chen W,Li S,Yang S,Zhang Y,Hu X,Qiu H,Wu J,Xu S,Chu T
Sat Oct 16 00:00:00 EDT 2021
Nox4 Promotes RANKL-Induced Autophagy and Osteoclastogenesis via Activating ROS/PERK/eIF-2α/ATF4 Pathway.
Frontiers in pharmacology
Sun J,Chen W,Li S,Yang S,Zhang Y,Hu X,Qiu H,Wu J,Xu S,Chu T
Sat Oct 16 00:00:00 EDT 2021
Dapagliflozin alleviates cardiac fibrosis through suppressing EndMT and fibroblast activation via AMPKα/TGF-β/Smad signalling in type 2 diabetic rats.
Journal of cellular and molecular medicine
Tian J,Zhang M,Suo M,Liu D,Wang X,Liu M,Pan J,Jin T,An F
Sun Aug 01 00:00:00 EDT 2021
Dapagliflozin alleviates cardiac fibrosis through suppressing EndMT and fibroblast activation via AMPKα/TGF-β/Smad signalling in type 2 diabetic rats.
Journal of cellular and molecular medicine
Tian J,Zhang M,Suo M,Liu D,Wang X,Liu M,Pan J,Jin T,An F
Sun Aug 01 00:00:00 EDT 2021
JI017, a Complex Herbal Medication, Induces Apoptosis via the Nox4-PERK-CHOP Axis in Ovarian Cancer Cells.
International journal of molecular sciences
Kim T,Ko SG
Fri Nov 12 00:00:00 EST 2021
Recombinant Human Growth Hormone Inhibits Lipotoxicity, Oxidative Stress, and Apoptosis in a Mouse Model of Diabetic Cardiomyopathy.
Oxidative medicine and cellular longevity
Pei Z,Wang X,Yang C,Dong M,Wang F
Mon Feb 28 00:00:00 EST 2022
Knockdown of Salusin-β Improves Cardiovascular Function in Myocardial Infarction-Induced Chronic Heart Failure Rats.
Oxidative medicine and cellular longevity
Xu Y,Pan Y,Wang X,Chen A,Tang X,Liu X,Han Y
Tue Jan 11 00:00:00 EST 2022
Role of oncogene PIM-1 in the development and progression of papillary thyroid carcinoma: Involvement of oxidative stress.
Molecular and cellular endocrinology
Wen QL,Yi HQ,Yang K,Yin CT,Yin WJ,Xiang FY,Bao M,Shuai J,Song YW,Ge MH,Zhu X
Mon Mar 01 00:00:00 EST 2021
Role of oncogene PIM-1 in the development and progression of papillary thyroid carcinoma: Involvement of oxidative stress.
Molecular and cellular endocrinology
Wen QL,Yi HQ,Yang K,Yin CT,Yin WJ,Xiang FY,Bao M,Shuai J,Song YW,Ge MH,Zhu X
Mon Mar 01 00:00:00 EST 2021
Effect of different exercise training intensities on age-related cardiac damage in male mice.
Aging
Pei Z,Yang C,Guo Y,Dong M,Wang F
Tue Sep 14 00:00:00 EDT 2021
Effect of different exercise training intensities on age-related cardiac damage in male mice.
Aging
Pei Z,Yang C,Guo Y,Dong M,Wang F
Tue Sep 14 00:00:00 EDT 2021
Sitagliptin Mitigates Total Body Irradiation-Induced Hematopoietic Injury in Mice.
Oxidative medicine and cellular longevity
Wang M,Dong Y,Wu J,Li H,Zhang J,Lu L,Zhang Y,Zhou Z,Fan S,Li D,Meng A
Fri May 14 00:00:00 EDT 2021
TXNIP deficiency mitigates podocyte apoptosis via restraining the activation of mTOR or p38 MAPK signaling in diabetic nephropathy.
Experimental cell research
Song S,Qiu D,Wang Y,Wei J,Wu H,Wu M,Wang S,Zhou X,Shi Y,Duan H
Sun Mar 15 00:00:00 EDT 2020
Punicalagin Protects Diabetic Nephropathy by Inhibiting Pyroptosis Based on TXNIP/NLRP3 Pathway.
Nutrients
An X,Zhang Y,Cao Y,Chen J,Qin H,Yang L
Fri May 22 00:00:00 EDT 2020
Cathelicidin Modulates Vascular Smooth Muscle Cell Phenotypic Switching through ROS/IL-6 Pathway.
Antioxidants (Basel, Switzerland)
Dong X,Wu D,Zhang Y,Jia L,Pan X,Sun J,Pan LL
Fri Jun 05 00:00:00 EDT 2020
A novel PPAR¿ ligand, PPZ023, overcomes radioresistance via ER stress and cell death in human non-small-cell lung cancer cells.
Experimental & molecular medicine
Kim TW,Hong DW,Kang CM,Hong SH
Thu Oct 01 00:00:00 EDT 2020
S-Allylmercaptocysteine Targets Nrf2 in Osteoarthritis Treatment Through NOX4/NF-κB Pathway.
Drug design, development and therapy
Yang G,Sun S,Wang J,Li W,Wang X,Yuan L,Li S
Tue Aug 24 00:00:00 EDT 2021
Cellular senescence and EMT crosstalk in bleomycin-induced pathogenesis of pulmonary fibrosis-an in vitro analysis.
Cell biology international
Muthuramalingam K,Cho M,Kim Y
Sat Feb 01 00:00:00 EST 2020
Ginkgetin Alleviates Inflammation, Oxidative Stress, and Apoptosis Induced by Hypoxia/Reoxygenation in H9C2 Cells via Caspase-3 Dependent Pathway.
BioMed research international
Liu X,Bian H,Dou QL,Huang XW,Tao WY,Liu WH,Li N,Zhang WW
Wed Mar 17 00:00:00 EDT 2021
Setanaxib as a Potent Hypoxia-specific Therapeutic Agent Against Liver Cancer.
Anticancer research
Owada S,Endo H,Okada C,Yoshida K,Shida Y,Tatemichi M
Tue Sep 01 00:00:00 EDT 2020
MiR155-5p Inhibits Cell Migration and Oxidative Stress in Vascular Smooth Muscle Cells of Spontaneously Hypertensive Rats.
Antioxidants (Basel, Switzerland)
Wu N,Ye C,Zheng F,Wan GW,Wu LL,Chen Q,Li YH,Kang YM,Zhu GQ
Sun Mar 01 00:00:00 EST 2020
Polydatin attenuates renal fibrosis in diabetic mice through regulating the Cx32-Nox4 signaling pathway.
Acta pharmacologica Sinica
Chen ZQ,Sun XH,Li XJ,Xu ZC,Yang Y,Lin ZY,Xiao HM,Zhang M,Quan SJ,Huang HQ
Tue Dec 01 00:00:00 EST 2020
Connexin32 ameliorates renal fibrosis in diabetic mice by promoting K48-linked NADPH oxidase 4 polyubiquitination and degradation.
British journal of pharmacology
Chen Z,Sun X,Chen Q,Lan T,Huang K,Xiao H,Lin Z,Yang Y,Liu P,Huang H
Wed Jan 01 00:00:00 EST 2020
Empagliflozin improves diabetic renal tubular injury by alleviating mitochondrial fission via AMPK/SP1/PGAM5 pathway.
Metabolism: clinical and experimental
Liu X,Xu C,Xu L,Li X,Sun H,Xue M,Li T,Yu X,Sun B,Chen L
Thu Oct 01 00:00:00 EDT 2020
S-Allylmercaptocysteine attenuates Bleomycin-induced pulmonary fibrosis in mice via suppressing TGF-β1/Smad and oxidative stress pathways.
International immunopharmacology
Li C,Sun X,Li A,Mo M,Zhao Z
Sat Feb 01 00:00:00 EST 2020
Induction of SPARC on Oxidative Stress, Inflammatory Phenotype Transformation, and Apoptosis of Human Brain Smooth Muscle Cells Via TGF-β1-NOX4 Pathway.
Journal of molecular neuroscience : MN
Tan X,Li T,Zhu S,Zhong W,Li F,Wang Y
Sun Nov 01 00:00:00 EDT 2020
FNDC5 Attenuates Oxidative Stress and NLRP3 Inflammasome Activation in Vascular Smooth Muscle Cells via Activating the AMPK-SIRT1 Signal Pathway.
Oxidative medicine and cellular longevity
Zhou B,Qiu Y,Wu N,Chen AD,Zhou H,Chen Q,Kang YM,Li YH,Zhu GQ
Mon Dec 21 00:00:00 EST 2020
DUOX2, a common modulator in preventive effects of monoamine-based antidepressants on water immersion restraint stress- and indomethacin- induced gastric mucosal damage.
European journal of pharmacology
Cao L,Li X,Xu R,Yao K,Yang W,Zhu H,Wang G,Zhang J
Fri Jun 05 00:00:00 EDT 2020
Morroniside Inhibits H2O2-Induced Podocyte Apoptosis by Down-Regulating NOX4 Expression Controlled by Autophagy In Vitro.
Frontiers in pharmacology
Gao X,Liu Y,Wang L,Sai N,Liu Y,Ni J
Thu Oct 22 00:00:00 EDT 2020
Small molecule inhibition of cyclic GMP-AMP synthase ameliorates sepsis-induced cardiac dysfunction in mice.
Life sciences
Xu Q,Xiong H,Zhu W,Liu Y,Du Y
Sun Nov 01 00:00:00 EDT 2020
Protective effect of ginsenoside Rg5 against kidney injury via inhibition of NLRP3 inflammasome activation and the MAPK signaling pathway in high-fat diet/streptozotocin-induced diabetic mice.
Pharmacological research
Zhu Y,Zhu C,Yang H,Deng J,Fan D
Fri May 01 00:00:00 EDT 2020
Protective effect of ginsenoside Rg5 against kidney injury via inhibition of NLRP3 inflammasome activation and the MAPK signaling pathway in high-fat diet/streptozotocin-induced diabetic mice.
Pharmacological research
Zhu Y,Zhu C,Yang H,Deng J,Fan D
Fri May 01 00:00:00 EDT 2020
Plumbagin attenuated oxygen-glucose deprivation/reoxygenation-induced injury in human SH-SY5Y cells by inhibiting NOX4-derived ROS-activated NLRP3 inflammasome.
Bioscience, biotechnology, and biochemistry
Zhang Q,Zhao S,Zheng W,Fu H,Wu T,Hu F
Wed Jan 01 00:00:00 EST 2020
Magnesium isoglycyrrhizinate alleviates fructose-induced liver oxidative stress and inflammatory injury through suppressing NOXs.
European journal of pharmacology
Yang YZ,Liu ZH,Wang SC,Zhang XQ,Xu HJ,Yang L,Kong LD
Tue Sep 15 00:00:00 EDT 2020
Magnesium isoglycyrrhizinate alleviates fructose-induced liver oxidative stress and inflammatory injury through suppressing NOXs.
European journal of pharmacology
Yang YZ,Liu ZH,Wang SC,Zhang XQ,Xu HJ,Yang L,Kong LD
Tue Sep 15 00:00:00 EDT 2020
Asiatic acid prevents renal fibrosis in UUO rats via promoting the production of 15d-PGJ2, an endogenous ligand of PPAR-γ.
Acta pharmacologica Sinica
Zhang ZH,He JQ,Zhao YY,Chen HC,Tan NH
Sun Mar 01 00:00:00 EST 2020
Novel crosstalk between Vps26a and Nox4 signaling during neurogenesis.
Cell death and differentiation
Choi SA,Kim YH,Park YH,Yang HJ,Jeong PS,Cha JJ,Yoon SB,Kim JS,Song BS,Lee JH,Sim BW,Huh JW,Song IS,Lee SR,Kim MK,Kim JM,Bae YS,Imakawa K,Kim SU,Chang KT
Sun Sep 01 00:00:00 EDT 2019
Novel crosstalk between Vps26a and Nox4 signaling during neurogenesis.
Cell death and differentiation
Choi SA,Kim YH,Park YH,Yang HJ,Jeong PS,Cha JJ,Yoon SB,Kim JS,Song BS,Lee JH,Sim BW,Huh JW,Song IS,Lee SR,Kim MK,Kim JM,Bae YS,Imakawa K,Kim SU,Chang KT
Sun Sep 01 00:00:00 EDT 2019
Novel crosstalk between Vps26a and Nox4 signaling during neurogenesis.
Cell death and differentiation
Choi SA,Kim YH,Park YH,Yang HJ,Jeong PS,Cha JJ,Yoon SB,Kim JS,Song BS,Lee JH,Sim BW,Huh JW,Song IS,Lee SR,Kim MK,Kim JM,Bae YS,Imakawa K,Kim SU,Chang KT
Sun Sep 01 00:00:00 EDT 2019
Novel crosstalk between Vps26a and Nox4 signaling during neurogenesis.
Cell death and differentiation
Choi SA,Kim YH,Park YH,Yang HJ,Jeong PS,Cha JJ,Yoon SB,Kim JS,Song BS,Lee JH,Sim BW,Huh JW,Song IS,Lee SR,Kim MK,Kim JM,Bae YS,Imakawa K,Kim SU,Chang KT
Sun Sep 01 00:00:00 EDT 2019
A Cross Talk Between BRG1 and Males Absent on the First Contributes to Reactive Oxygen Species Production in a Mouse Model of Nonalcoholic Steatohepatitis.
Antioxidants & redox signaling
Liu L,Hong W,Li M,Ren H,Wang J,Xu H,Shi X,Xu Y
Sat Apr 20 00:00:00 EDT 2019
A Cross Talk Between BRG1 and Males Absent on the First Contributes to Reactive Oxygen Species Production in a Mouse Model of Nonalcoholic Steatohepatitis.
Antioxidants & redox signaling
Liu L,Hong W,Li M,Ren H,Wang J,Xu H,Shi X,Xu Y
Sat Apr 20 00:00:00 EDT 2019
Potential mechanisms underlying the protective effects of salvianic acid A against atherosclerosis in vivo and vitro.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Song Q,Zhang Y,Han X,Zhang Y,Zhang X,Gao Y,Zhang J,Chu L,Zhao S
Tue Jan 01 00:00:00 EST 2019
Potential mechanisms underlying the protective effects of salvianic acid A against atherosclerosis in vivo and vitro.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Song Q,Zhang Y,Han X,Zhang Y,Zhang X,Gao Y,Zhang J,Chu L,Zhao S
Tue Jan 01 00:00:00 EST 2019
Sensory plasticity of carotid body is correlated with oxidative stress in paraventricular nucleus during chronic intermittent hypoxia.
Journal of cellular physiology
Liu P,Zhang HM,Hu K,Zhou XF,Tang S
Thu Aug 01 00:00:00 EDT 2019
Sensory plasticity of carotid body is correlated with oxidative stress in paraventricular nucleus during chronic intermittent hypoxia.
Journal of cellular physiology
Liu P,Zhang HM,Hu K,Zhou XF,Tang S
Thu Aug 01 00:00:00 EDT 2019
Catalpol Inhibits Homocysteine-induced Oxidation and Inflammation via Inhibiting Nox4/NF-κB and GRP78/PERK Pathways in Human Aorta Endothelial Cells.
Inflammation
Hu H,Wang C,Jin Y,Meng Q,Liu Q,Liu Z,Liu K,Liu X,Sun H
Fri Feb 01 00:00:00 EST 2019
Catalpol Inhibits Homocysteine-induced Oxidation and Inflammation via Inhibiting Nox4/NF-κB and GRP78/PERK Pathways in Human Aorta Endothelial Cells.
Inflammation
Hu H,Wang C,Jin Y,Meng Q,Liu Q,Liu Z,Liu K,Liu X,Sun H
Fri Feb 01 00:00:00 EST 2019
Allicin attenuated chronic social defeat stress induced depressive-like behaviors through suppression of NLRP3 inflammasome.
Metabolic brain disease
Gao W,Wang W,Liu G,Zhang J,Yang J,Deng Z
Fri Feb 01 00:00:00 EST 2019
Allicin attenuated chronic social defeat stress induced depressive-like behaviors through suppression of NLRP3 inflammasome.
Metabolic brain disease
Gao W,Wang W,Liu G,Zhang J,Yang J,Deng Z
Fri Feb 01 00:00:00 EST 2019
SRT1720 retards renal fibrosis via inhibition of HIF1α /GLUT1 in diabetic nephropathy.
The Journal of endocrinology
Han W,Wang C,Yang Z,Mu L,Wu M,Chen N,Du C,Duan H,Shi Y
Fri Feb 01 00:00:00 EST 2019
Systemic Outcomes of (Pyr1)-Apelin-13 Infusion at Mid-Late Pregnancy in a Rat Model with Preeclamptic Features.
Scientific reports
Yamaleyeva LM,Brosnihan KB,Elsangeedy E,McGee C,Shi S,Caudell D,Miller C,Varagic J,Bader M,Dechend R,Shaltout HA
Wed Jun 12 00:00:00 EDT 2019
Protective Effects of 18β-Glycyrrhetinic Acid on Monocrotaline-Induced Pulmonary Arterial Hypertension in Rats.
Frontiers in pharmacology
Zhang M,Chang Z,Zhao F,Zhang P,Hao YJ,Yan L,Liu N,Wang JL,Bo L,Ma P,Zhou W,Ma X,Xu QB,Zhou R
Thu Oct 01 00:00:00 EDT 2020
Inducible nitric oxide synthase contributes to insulin resistance and cardiac dysfunction after burn injury in mice.
Life sciences
Guo Y,You Y,Lv D,Yan J,Shang FF,Wang X,Zhang C,Fan Q,Luo S
Sun Dec 15 00:00:00 EST 2019
Salvianolic Acid A Protects the Kidney against Oxidative Stress by Activating the Akt/GSK-3β/Nrf2 Signaling Pathway and Inhibiting the NF-κB Signaling Pathway in 5/6 Nephrectomized Rats.
Oxidative medicine and cellular longevity
Zhang HF,Wang JH,Wang YL,Gao C,Gu YT,Huang J,Wang JH,Zhang Z
Thu Aug 08 00:00:00 EDT 2019
MicroRNA-24 inhibits the oxidative stress induced by vascular injury by activating the Nrf2/Ho-1 signaling pathway.
Atherosclerosis
Zhang J,Cai W,Fan Z,Yang C,Wang W,Xiong M,Ma C,Yang J
Fri Nov 01 00:00:00 EDT 2019
MARVELD1 interacting with catalase regulates reactive oxygen species metabolism and mediates the sensitivity to chemotherapeutic drugs in epithelial tumors of the reproductive system.
Molecular carcinogenesis
Zhang C,Han F,Shi M,Sun H,Li Y,Ci Y,Yao Y,Dou P,Akhtar ML,Nie H,He J,Li Y
Thu Aug 01 00:00:00 EDT 2019
Glutathione prevents chronic oscillating glucose intake-induced β-cell dedifferentiation and failure.
Cell death & disease
Zhang J,An H,Ni K,Chen B,Li H,Li Y,Sheng G,Zhou C,Xie M,Chen S,Zhou T,Yang G,Chen X,Wu G,Jin S,Li M
Thu Apr 11 00:00:00 EDT 2019
Disocin prevents postmenopausal atherosclerosis in ovariectomized LDLR-/- mice through a PGC-1α/ERα pathway leading to promotion of autophagy and inhibition of oxidative stress, inflammation and apoptosis.
Pharmacological research
Yang Q,Wang C,Jin Y,Ma X,Xie T,Wang J,Liu K,Sun H
Tue Oct 01 00:00:00 EDT 2019
Vitamin D receptor activation regulates microglia polarization and oxidative stress in spontaneously hypertensive rats and angiotensin II-exposed microglial cells: Role of renin-angiotensin system.
Redox biology
Cui C,Xu P,Li G,Qiao Y,Han W,Geng C,Liao D,Yang M,Chen D,Jiang P
Sun Sep 01 00:00:00 EDT 2019
Spatial oxidation of L-plastin downmodulates actin-based functions of tumor cells.
Nature communications
Balta E,Hardt R,Liang J,Kirchgessner H,Orlik C,Jahraus B,Hillmer S,Meuer S,Hübner K,Wabnitz GH,Samstag Y
Mon Sep 09 00:00:00 EDT 2019
Costunolide inhibits pulmonary fibrosis via regulating NF-kB and TGF-β1/Smad2/Nrf2-NOX4 signaling pathways.
Biochemical and biophysical research communications
Liu B,Rong Y,Sun D,Li W,Chen H,Cao B,Wang T
Tue Mar 05 00:00:00 EST 2019
Astragaloside IV alleviates doxorubicin induced cardiomyopathy by inhibiting NADPH oxidase derived oxidative stress.
European journal of pharmacology
Lin J,Fang L,Li H,Li Z,Lyu L,Wang H,Xiao J
Sun Sep 15 00:00:00 EDT 2019
Thioredoxin-interacting protein deficiency alleviates phenotypic alterations of podocytes via inhibition of mTOR activation in diabetic nephropathy.
Journal of cellular physiology
Song S,Qiu D,Shi Y,Wang S,Zhou X,Chen N,Wei J,Wu M,Wu H,Duan H
Sun Sep 01 00:00:00 EDT 2019
Thioredoxin-interacting protein deficiency alleviates phenotypic alterations of podocytes via inhibition of mTOR activation in diabetic nephropathy.
Journal of cellular physiology
Song S,Qiu D,Shi Y,Wang S,Zhou X,Chen N,Wei J,Wu M,Wu H,Duan H
Sun Sep 01 00:00:00 EDT 2019
Thioredoxin-interacting protein deficiency alleviates phenotypic alterations of podocytes via inhibition of mTOR activation in diabetic nephropathy.
Journal of cellular physiology
Song S,Qiu D,Shi Y,Wang S,Zhou X,Chen N,Wei J,Wu M,Wu H,Duan H
Sun Sep 01 00:00:00 EDT 2019
Thioredoxin-interacting protein deficiency alleviates phenotypic alterations of podocytes via inhibition of mTOR activation in diabetic nephropathy.
Journal of cellular physiology
Song S,Qiu D,Shi Y,Wang S,Zhou X,Chen N,Wei J,Wu M,Wu H,Duan H
Sun Sep 01 00:00:00 EDT 2019
Effects of Dexmedetomidine on the RhoA /ROCK/ Nox4 Signaling Pathway in Renal Fibrosis of Diabetic Rats.
Open medicine (Warsaw, Poland)
Jihua C,Cai C,Xubin B,Yue Y
Mon Apr 11 00:00:00 EDT 2022
Calcium Release from Endoplasmic Reticulum Involves Calmodulin-Mediated NADPH Oxidase-Derived Reactive Oxygen Species Production in Endothelial Cells.
International journal of molecular sciences
Sakurada R,Odagiri K,Hakamata A,Kamiya C,Wei J,Watanabe H
Tue Apr 02 00:00:00 EDT 2019
Empagliflozin prevents cardiomyopathy via sGC-cGMP-PKG pathway in type 2 diabetes mice.
Clinical science (London, England : 1979)
Xue M,Li T,Wang Y,Chang Y,Cheng Y,Lu Y,Liu X,Xu L,Li X,Yu X,Sun B,Chen L
Thu Aug 15 00:00:00 EDT 2019
Neuroprotective Effect of SCM-198 through Stabilizing Endothelial Cell Function.
Oxidative medicine and cellular longevity
Zhang QY,Wang ZJ,Miao L,Wang Y,Chang LL,Guo W,Zhu YZ
Mon May 04 00:00:00 EDT 2020
Neuroprotective Effect of SCM-198 through Stabilizing Endothelial Cell Function.
Oxidative medicine and cellular longevity
Zhang QY,Wang ZJ,Miao L,Wang Y,Chang LL,Guo W,Zhu YZ
Mon May 04 00:00:00 EDT 2020
Synergistically dual-functional nano eye-drops for simultaneous anti-inflammatory and anti-oxidative treatment of dry eye disease.
Nanoscale
Li YJ,Luo LJ,Harroun SG,Wei SC,Unnikrishnan B,Chang HT,Huang YF,Lai JY,Huang CC
Thu Mar 21 00:00:00 EDT 2019
Growth differentiation factor 11 is involved in isoproterenol‑induced heart failure.
Molecular medicine reports
Zhang XJ,Tan H,Shi ZF,Li N,Jia Y,Hao Z
Wed May 01 00:00:00 EDT 2019
BCL6 Attenuates Proliferation and Oxidative Stress of Vascular Smooth Muscle Cells in Hypertension.
Oxidative medicine and cellular longevity
Chen D,Zang YH,Qiu Y,Zhang F,Chen AD,Wang JJ,Chen Q,Li YH,Kang YM,Zhu GQ
Mon Mar 25 00:00:00 EDT 2019
Allicin ameliorates obesity comorbid depressive-like behaviors: involvement of the oxidative stress, mitochondrial function, autophagy, insulin resistance and NOX/Nrf2 imbalance in mice.
Metabolic brain disease
Gao W,Wang W,Zhang J,Deng P,Hu J,Yang J,Deng Z
Tue Oct 01 00:00:00 EDT 2019
Allicin ameliorates obesity comorbid depressive-like behaviors: involvement of the oxidative stress, mitochondrial function, autophagy, insulin resistance and NOX/Nrf2 imbalance in mice.
Metabolic brain disease
Gao W,Wang W,Zhang J,Deng P,Hu J,Yang J,Deng Z
Tue Oct 01 00:00:00 EDT 2019
Megakaryocytic Leukemia 1 Bridges Epigenetic Activation of NADPH Oxidase in Macrophages to Cardiac Ischemia-Reperfusion Injury.
Circulation
Yu L,Yang G,Zhang X,Wang P,Weng X,Yang Y,Li Z,Fang M,Xu Y,Sun A,Ge J
Tue Dec 11 00:00:00 EST 2018
Megakaryocytic Leukemia 1 Bridges Epigenetic Activation of NADPH Oxidase in Macrophages to Cardiac Ischemia-Reperfusion Injury.
Circulation
Yu L,Yang G,Zhang X,Wang P,Weng X,Yang Y,Li Z,Fang M,Xu Y,Sun A,Ge J
Tue Dec 11 00:00:00 EST 2018
Catalpol ameliorates hepatic insulin resistance in type 2 diabetes through acting on AMPK/NOX4/PI3K/AKT pathway.
Pharmacological research
Yan J,Wang C,Jin Y,Meng Q,Liu Q,Liu Z,Liu K,Sun H
Sun Apr 01 00:00:00 EDT 2018
Catalpol ameliorates hepatic insulin resistance in type 2 diabetes through acting on AMPK/NOX4/PI3K/AKT pathway.
Pharmacological research
Yan J,Wang C,Jin Y,Meng Q,Liu Q,Liu Z,Liu K,Sun H
Sun Apr 01 00:00:00 EDT 2018
MCUR1-Mediated Mitochondrial Calcium Signaling Facilitates Cell Survival of Hepatocellular Carcinoma via Reactive Oxygen Species-Dependent P53 Degradation.
Antioxidants & redox signaling
Ren T,Wang J,Zhang H,Yuan P,Zhu J,Wu Y,Huang Q,Guo X,Zhang J,Ji L,Li J,Zhang H,Yang H,Xing J
Fri Apr 20 00:00:00 EDT 2018
MCUR1-Mediated Mitochondrial Calcium Signaling Facilitates Cell Survival of Hepatocellular Carcinoma via Reactive Oxygen Species-Dependent P53 Degradation.
Antioxidants & redox signaling
Ren T,Wang J,Zhang H,Yuan P,Zhu J,Wu Y,Huang Q,Guo X,Zhang J,Ji L,Li J,Zhang H,Yang H,Xing J
Fri Apr 20 00:00:00 EDT 2018
CAPE-pNO2 attenuates diabetic cardiomyopathy through the NOX4/NF-κB pathway in STZ-induced diabetic mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Fan L,Xiao Q,Zhang L,Wang X,Huang Q,Li S,Zhao X,Li Z
Sat Dec 01 00:00:00 EST 2018
CAPE-pNO2 attenuates diabetic cardiomyopathy through the NOX4/NF-κB pathway in STZ-induced diabetic mice.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Fan L,Xiao Q,Zhang L,Wang X,Huang Q,Li S,Zhao X,Li Z
Sat Dec 01 00:00:00 EST 2018
Downregulated Expression of Solute Carrier Family 26 Member 6 in NRK-52E Cells Attenuates Oxalate-Induced Intracellular Oxidative Stress.
Oxidative medicine and cellular longevity
Jiang H,Gao X,Gong J,Yang Q,Lan R,Wang T,Liu J,Yin C,Wang S,Liu Z
Mon Nov 26 00:00:00 EST 2018
Downregulated Expression of Solute Carrier Family 26 Member 6 in NRK-52E Cells Attenuates Oxalate-Induced Intracellular Oxidative Stress.
Oxidative medicine and cellular longevity
Jiang H,Gao X,Gong J,Yang Q,Lan R,Wang T,Liu J,Yin C,Wang S,Liu Z
Mon Nov 26 00:00:00 EST 2018
The Kidney Injury Induced by Short-Term PM2.5 Exposure and the Prophylactic Treatment of Essential Oils in BALB/c Mice.
Oxidative medicine and cellular longevity
Zhang Y,Li Q,Fang M,Ma Y,Liu N,Yan X,Zhou J,Li F
Tue Dec 11 00:00:00 EST 2018
The Kidney Injury Induced by Short-Term PM2.5 Exposure and the Prophylactic Treatment of Essential Oils in BALB/c Mice.
Oxidative medicine and cellular longevity
Zhang Y,Li Q,Fang M,Ma Y,Liu N,Yan X,Zhou J,Li F
Tue Dec 11 00:00:00 EST 2018
BRG1 regulates NOX gene transcription in endothelial cells and contributes to cardiac ischemia-reperfusion injury.
Biochimica et biophysica acta. Molecular basis of disease
Li Z,Zhang X,Liu S,Zeng S,Yu L,Yang G,Guo J,Xu Y
Mon Oct 01 00:00:00 EDT 2018
BRG1 regulates NOX gene transcription in endothelial cells and contributes to cardiac ischemia-reperfusion injury.
Biochimica et biophysica acta. Molecular basis of disease
Li Z,Zhang X,Liu S,Zeng S,Yu L,Yang G,Guo J,Xu Y
Mon Oct 01 00:00:00 EDT 2018
Myocardin-related transcription factor A (MRTF-A) contributes to acute kidney injury by regulating macrophage ROS production.
Biochimica et biophysica acta. Molecular basis of disease
Liu L,Wu X,Xu H,Yu L,Zhang X,Li L,Jin J,Zhang T,Xu Y
Mon Oct 01 00:00:00 EDT 2018
Myocardin-related transcription factor A (MRTF-A) contributes to acute kidney injury by regulating macrophage ROS production.
Biochimica et biophysica acta. Molecular basis of disease
Liu L,Wu X,Xu H,Yu L,Zhang X,Li L,Jin J,Zhang T,Xu Y
Mon Oct 01 00:00:00 EDT 2018
Resveratrol attenuates hyperproliferation of vascular smooth muscle cells from spontaneously hypertensive rats: Role of ROS and ROS-mediated cell signaling.
Vascular pharmacology
Almajdoob S,Hossain E,Anand-Srivastava MB
Thu Feb 01 00:00:00 EST 2018
Resveratrol attenuates hyperproliferation of vascular smooth muscle cells from spontaneously hypertensive rats: Role of ROS and ROS-mediated cell signaling.
Vascular pharmacology
Almajdoob S,Hossain E,Anand-Srivastava MB
Thu Feb 01 00:00:00 EST 2018
A novel Gallic acid derivative attenuates BLM-induced pulmonary fibrosis in mice.
International immunopharmacology
Rong Y,Cao B,Liu B,Li W,Chen Y,Chen H,Liu Y,Liu T
Thu Nov 01 00:00:00 EDT 2018
A novel Gallic acid derivative attenuates BLM-induced pulmonary fibrosis in mice.
International immunopharmacology
Rong Y,Cao B,Liu B,Li W,Chen Y,Chen H,Liu Y,Liu T
Thu Nov 01 00:00:00 EDT 2018
A novel Gallic acid derivative attenuates BLM-induced pulmonary fibrosis in mice.
International immunopharmacology
Rong Y,Cao B,Liu B,Li W,Chen Y,Chen H,Liu Y,Liu T
Thu Nov 01 00:00:00 EDT 2018
A novel Gallic acid derivative attenuates BLM-induced pulmonary fibrosis in mice.
International immunopharmacology
Rong Y,Cao B,Liu B,Li W,Chen Y,Chen H,Liu Y,Liu T
Thu Nov 01 00:00:00 EDT 2018
Dark tea extract mitigates hematopoietic radiation injury with antioxidative activity.
Journal of radiation research
Long W,Zhang G,Dong Y,Li D
Sun Jul 01 00:00:00 EDT 2018
Dark tea extract mitigates hematopoietic radiation injury with antioxidative activity.
Journal of radiation research
Long W,Zhang G,Dong Y,Li D
Sun Jul 01 00:00:00 EDT 2018
Alpha-Lipoic Acid Protects Human Aortic Endothelial Cells Against H2O2-Induced Injury and Inhibits Atherosclerosis in Ovariectomized Low Density Lipoprotein Receptor Knock-Out Mice.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Shen D,Tian L,Shen T,Sun H,Liu P
Fri Aug 17 00:00:00 EDT 2018
Alpha-Lipoic Acid Protects Human Aortic Endothelial Cells Against H2O2-Induced Injury and Inhibits Atherosclerosis in Ovariectomized Low Density Lipoprotein Receptor Knock-Out Mice.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Shen D,Tian L,Shen T,Sun H,Liu P
Fri Aug 17 00:00:00 EDT 2018
Alpha-Lipoic Acid Protects Human Aortic Endothelial Cells Against H2O2-Induced Injury and Inhibits Atherosclerosis in Ovariectomized Low Density Lipoprotein Receptor Knock-Out Mice.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Shen D,Tian L,Shen T,Sun H,Liu P
Fri Aug 17 00:00:00 EDT 2018
Alpha-Lipoic Acid Protects Human Aortic Endothelial Cells Against H2O2-Induced Injury and Inhibits Atherosclerosis in Ovariectomized Low Density Lipoprotein Receptor Knock-Out Mice.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Shen D,Tian L,Shen T,Sun H,Liu P
Fri Aug 17 00:00:00 EDT 2018
Sodium Citrate Increases Expression and Flux of Mg2+ Transport Carriers Mediated by Activation of MEK/ERK/c-Fos Pathway in Renal Tubular Epithelial Cells.
Nutrients
Takashina Y,Manabe A,Hasegawa H,Matsunaga T,Endo S,Ikari A
Thu Sep 20 00:00:00 EDT 2018
Sodium Citrate Increases Expression and Flux of Mg2+ Transport Carriers Mediated by Activation of MEK/ERK/c-Fos Pathway in Renal Tubular Epithelial Cells.
Nutrients
Takashina Y,Manabe A,Hasegawa H,Matsunaga T,Endo S,Ikari A
Thu Sep 20 00:00:00 EDT 2018
Sodium Citrate Increases Expression and Flux of Mg2+ Transport Carriers Mediated by Activation of MEK/ERK/c-Fos Pathway in Renal Tubular Epithelial Cells.
Nutrients
Takashina Y,Manabe A,Hasegawa H,Matsunaga T,Endo S,Ikari A
Thu Sep 20 00:00:00 EDT 2018
Sodium Citrate Increases Expression and Flux of Mg2+ Transport Carriers Mediated by Activation of MEK/ERK/c-Fos Pathway in Renal Tubular Epithelial Cells.
Nutrients
Takashina Y,Manabe A,Hasegawa H,Matsunaga T,Endo S,Ikari A
Thu Sep 20 00:00:00 EDT 2018
Azithromycin attenuates myofibroblast differentiation and lung fibrosis development through proteasomal degradation of NOX4.
Autophagy
Tsubouchi K,Araya J,Minagawa S,Hara H,Ichikawa A,Saito N,Kadota T,Sato N,Yoshida M,Kurita Y,Kobayashi K,Ito S,Fujita Y,Utsumi H,Yanagisawa H,Hashimoto M,Wakui H,Yoshii Y,Ishikawa T,Numata T,Kaneko Y,Asano H,Yamashita M,Odaka M,Morikawa T,Nakayama K,Nakanishi Y,Kuwano K
Thu Aug 03 00:00:00 EDT 2017
Azithromycin attenuates myofibroblast differentiation and lung fibrosis development through proteasomal degradation of NOX4.
Autophagy
Tsubouchi K,Araya J,Minagawa S,Hara H,Ichikawa A,Saito N,Kadota T,Sato N,Yoshida M,Kurita Y,Kobayashi K,Ito S,Fujita Y,Utsumi H,Yanagisawa H,Hashimoto M,Wakui H,Yoshii Y,Ishikawa T,Numata T,Kaneko Y,Asano H,Yamashita M,Odaka M,Morikawa T,Nakayama K,Nakanishi Y,Kuwano K
Thu Aug 03 00:00:00 EDT 2017
Novel Therapeutic Effects of Leonurine On Ischemic Stroke: New Mechanisms of BBB Integrity.
Oxidative medicine and cellular longevity
Zhang QY,Wang ZJ,Sun DM,Wang Y,Xu P,Wu WJ,Liu XH,Zhu YZ
Mon May 07 00:00:00 EDT 2018
Novel Therapeutic Effects of Leonurine On Ischemic Stroke: New Mechanisms of BBB Integrity.
Oxidative medicine and cellular longevity
Zhang QY,Wang ZJ,Sun DM,Wang Y,Xu P,Wu WJ,Liu XH,Zhu YZ
Mon May 07 00:00:00 EDT 2018
Phosphocreatine protects endothelial cells from Methylglyoxal induced oxidative stress and apoptosis via the regulation of PI3K/Akt/eNOS and NF-κB pathway.
Vascular pharmacology
Chu P,Han G,Ahsan A,Sun Z,Liu S,Zhang Z,Sun B,Song Y,Lin Y,Peng J,Tang Z
Sat Apr 01 00:00:00 EDT 2017
Phosphocreatine protects endothelial cells from Methylglyoxal induced oxidative stress and apoptosis via the regulation of PI3K/Akt/eNOS and NF-κB pathway.
Vascular pharmacology
Chu P,Han G,Ahsan A,Sun Z,Liu S,Zhang Z,Sun B,Song Y,Lin Y,Peng J,Tang Z
Sat Apr 01 00:00:00 EDT 2017
Synergistic protection of Schizandrin B and Glycyrrhizic acid against bleomycin-induced pulmonary fibrosis by inhibiting TGF-β1/Smad2 pathways and overexpression of NOX4.
International immunopharmacology
Zhang D,Liu B,Cao B,Wei F,Yu X,Li GF,Chen H,Wei LQ,Wang PL
Sat Jul 01 00:00:00 EDT 2017
Synergistic protection of Schizandrin B and Glycyrrhizic acid against bleomycin-induced pulmonary fibrosis by inhibiting TGF-β1/Smad2 pathways and overexpression of NOX4.
International immunopharmacology
Zhang D,Liu B,Cao B,Wei F,Yu X,Li GF,Chen H,Wei LQ,Wang PL
Sat Jul 01 00:00:00 EDT 2017
Nox4 is involved in high glucose-induced apoptosis in renal tubular epithelial cells via Notch pathway.
Molecular medicine reports
Yao M,Gao F,Wang X,Shi Y,Liu S,Duan H
Thu Jun 01 00:00:00 EDT 2017
Nox4 is involved in high glucose-induced apoptosis in renal tubular epithelial cells via Notch pathway.
Molecular medicine reports
Yao M,Gao F,Wang X,Shi Y,Liu S,Duan H
Thu Jun 01 00:00:00 EDT 2017
The osteogenesis-promoting effects of alpha-lipoic acid against glucocorticoid-induced osteoporosis through the NOX4, NF-kappaB, JNK and PI3K/AKT pathways.
Scientific reports
Lu SY,Wang CY,Jin Y,Meng Q,Liu Q,Liu ZH,Liu KX,Sun HJ,Liu MZ
Tue Jun 13 00:00:00 EDT 2017
The osteogenesis-promoting effects of alpha-lipoic acid against glucocorticoid-induced osteoporosis through the NOX4, NF-kappaB, JNK and PI3K/AKT pathways.
Scientific reports
Lu SY,Wang CY,Jin Y,Meng Q,Liu Q,Liu ZH,Liu KX,Sun HJ,Liu MZ
Tue Jun 13 00:00:00 EDT 2017
Nephroprotective Effects of Saponins from Leaves of Panax quinquefolius against Cisplatin-Induced Acute Kidney Injury.
International journal of molecular sciences
Ma ZN,Li YZ,Li W,Yan XT,Yang G,Zhang J,Zhao LC,Yang LM
Thu Jul 13 00:00:00 EDT 2017
Nephroprotective Effects of Saponins from Leaves of Panax quinquefolius against Cisplatin-Induced Acute Kidney Injury.
International journal of molecular sciences
Ma ZN,Li YZ,Li W,Yan XT,Yang G,Zhang J,Zhao LC,Yang LM
Thu Jul 13 00:00:00 EDT 2017
Photoreceptor oxidative stress in hyperoxia-induced proliferative retinopathy accelerates rd8 degeneration.
PloS one
Lajko M,Cardona HJ,Taylor JM,Farrow KN,Fawzi AA
Fri Oct 06 00:00:00 EDT 2017
Photoreceptor oxidative stress in hyperoxia-induced proliferative retinopathy accelerates rd8 degeneration.
PloS one
Lajko M,Cardona HJ,Taylor JM,Farrow KN,Fawzi AA
Fri Oct 06 00:00:00 EDT 2017
Hydrogen Sulfide Improves Endothelial Dysfunction via Downregulating BMP4/COX-2 Pathway in Rats with Hypertension.
Oxidative medicine and cellular longevity
Xiao L,Dong JH,Jin S,Xue HM,Guo Q,Teng X,Wu YM
Mon Mar 13 00:00:00 EDT 2017
Hydrogen Sulfide Improves Endothelial Dysfunction via Downregulating BMP4/COX-2 Pathway in Rats with Hypertension.
Oxidative medicine and cellular longevity
Xiao L,Dong JH,Jin S,Xue HM,Guo Q,Teng X,Wu YM
Mon Mar 13 00:00:00 EDT 2017
Salvianolic acid B protects against paraquat-induced pulmonary injury by mediating Nrf2/Nox4 redox balance and TGF-β1/Smad3 signaling.
Toxicology and applied pharmacology
Liu B,Cao B,Zhang D,Xiao N,Chen H,Li GQ,Peng SC,Wei LQ
Sat Oct 15 00:00:00 EDT 2016
Salvianolic acid B protects against paraquat-induced pulmonary injury by mediating Nrf2/Nox4 redox balance and TGF-β1/Smad3 signaling.
Toxicology and applied pharmacology
Liu B,Cao B,Zhang D,Xiao N,Chen H,Li GQ,Peng SC,Wei LQ
Sat Oct 15 00:00:00 EDT 2016
Mixed Aqueous Extract of Salvia Miltiorrhiza Reduces Blood Pressure through Inhibition of Vascular Remodelling and Oxidative Stress in Spontaneously Hypertensive Rats.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Zhang J,An SJ,Fu JQ,Liu P,Shao TM,Li M,Li X,Jiao Z,Chai XQ
Mon Feb 13 00:00:00 EST 2017
Mixed Aqueous Extract of Salvia Miltiorrhiza Reduces Blood Pressure through Inhibition of Vascular Remodelling and Oxidative Stress in Spontaneously Hypertensive Rats.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Zhang J,An SJ,Fu JQ,Liu P,Shao TM,Li M,Li X,Jiao Z,Chai XQ
Mon Feb 13 00:00:00 EST 2017
Mixed Aqueous Extract of Salvia Miltiorrhiza Reduces Blood Pressure through Inhibition of Vascular Remodelling and Oxidative Stress in Spontaneously Hypertensive Rats.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Zhang J,An SJ,Fu JQ,Liu P,Shao TM,Li M,Li X,Jiao Z,Chai XQ
Mon Feb 13 00:00:00 EST 2017
Mixed Aqueous Extract of Salvia Miltiorrhiza Reduces Blood Pressure through Inhibition of Vascular Remodelling and Oxidative Stress in Spontaneously Hypertensive Rats.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
Zhang J,An SJ,Fu JQ,Liu P,Shao TM,Li M,Li X,Jiao Z,Chai XQ
Mon Feb 13 00:00:00 EST 2017
The Protective Effects of 5-Methoxytryptamine-α-lipoic Acid on Ionizing Radiation-Induced Hematopoietic Injury.
International journal of molecular sciences
Li D,Tian Z,Tang W,Zhang J,Lu L,Sun Z,Zhou Z,Fan F
Tue Jun 14 00:00:00 EDT 2016
The Protective Effects of 5-Methoxytryptamine-α-lipoic Acid on Ionizing Radiation-Induced Hematopoietic Injury.
International journal of molecular sciences
Li D,Tian Z,Tang W,Zhang J,Lu L,Sun Z,Zhou Z,Fan F
Tue Jun 14 00:00:00 EDT 2016
ROS-mediated Different Homeostasis of Murine Corneal Epithelial Progenitor Cell Line under Oxidative Stress.
Scientific reports
Zhou J,Ge L,Jia C,Zheng X,Cui H,Zong R,Bao X,Yin Y,Ma JX,Li W,Liu Z,Zhou Y
Wed Nov 02 00:00:00 EDT 2016
ROS-mediated Different Homeostasis of Murine Corneal Epithelial Progenitor Cell Line under Oxidative Stress.
Scientific reports
Zhou J,Ge L,Jia C,Zheng X,Cui H,Zong R,Bao X,Yin Y,Ma JX,Li W,Liu Z,Zhou Y
Wed Nov 02 00:00:00 EDT 2016
ROS-mediated Different Homeostasis of Murine Corneal Epithelial Progenitor Cell Line under Oxidative Stress.
Scientific reports
Zhou J,Ge L,Jia C,Zheng X,Cui H,Zong R,Bao X,Yin Y,Ma JX,Li W,Liu Z,Zhou Y
Wed Nov 02 00:00:00 EDT 2016
ROS-mediated Different Homeostasis of Murine Corneal Epithelial Progenitor Cell Line under Oxidative Stress.
Scientific reports
Zhou J,Ge L,Jia C,Zheng X,Cui H,Zong R,Bao X,Yin Y,Ma JX,Li W,Liu Z,Zhou Y
Wed Nov 02 00:00:00 EDT 2016
Metformin ameliorates ionizing irradiation-induced long-term hematopoietic stem cell injury in mice.
Free radical biology & medicine
Xu G,Wu H,Zhang J,Li D,Wang Y,Wang Y,Zhang H,Lu L,Li C,Huang S,Xing Y,Zhou D,Meng A
Thu Oct 01 00:00:00 EDT 2015
Metformin ameliorates ionizing irradiation-induced long-term hematopoietic stem cell injury in mice.
Free radical biology & medicine
Xu G,Wu H,Zhang J,Li D,Wang Y,Wang Y,Zhang H,Lu L,Li C,Huang S,Xing Y,Zhou D,Meng A
Thu Oct 01 00:00:00 EDT 2015
Alpha-Lipoic Acid Promotes Osteoblastic Formation in H2O2 -Treated MC3T3-E1 Cells and Prevents Bone Loss in Ovariectomized Rats.
Journal of cellular physiology
Fu C,Xu D,Wang CY,Jin Y,Liu Q,Meng Q,Liu KX,Sun HJ,Liu MZ
Tue Sep 01 00:00:00 EDT 2015
Alpha-Lipoic Acid Promotes Osteoblastic Formation in H2O2 -Treated MC3T3-E1 Cells and Prevents Bone Loss in Ovariectomized Rats.
Journal of cellular physiology
Fu C,Xu D,Wang CY,Jin Y,Liu Q,Meng Q,Liu KX,Sun HJ,Liu MZ
Tue Sep 01 00:00:00 EDT 2015
Rhizoma Dioscoreae Nipponicae polysaccharides protect HUVECs from H2O2-induced injury by regulating PPARγ factor and the NADPH oxidase/ROS-NF-κB signal pathway.
Toxicology letters
Jin Y,Liu K,Peng J,Wang C,Kang L,Chang N,Sun H
Mon Jan 05 00:00:00 EST 2015
Rhizoma Dioscoreae Nipponicae polysaccharides protect HUVECs from H2O2-induced injury by regulating PPARγ factor and the NADPH oxidase/ROS-NF-κB signal pathway.
Toxicology letters
Jin Y,Liu K,Peng J,Wang C,Kang L,Chang N,Sun H
Mon Jan 05 00:00:00 EST 2015
PKCα promotes generation of reactive oxygen species via DUOX2 in hepatocellular carcinoma.
Biochemical and biophysical research communications
Wang J,Shao M,Liu M,Peng P,Li L,Wu W,Wang L,Duan F,Zhang M,Song S,Jia D,Ruan Y,Gu J
Fri Aug 07 00:00:00 EDT 2015
PKCα promotes generation of reactive oxygen species via DUOX2 in hepatocellular carcinoma.
Biochemical and biophysical research communications
Wang J,Shao M,Liu M,Peng P,Li L,Wu W,Wang L,Duan F,Zhang M,Song S,Jia D,Ruan Y,Gu J
Fri Aug 07 00:00:00 EDT 2015
{2-[1-(3-Methoxycarbonylmethyl-1H-indol-2-yl)-1-methyl-ethyl]-1H-indol-3-yl}-acetic Acid Methyl Ester Inhibited Hepatocellular Carcinoma Growth in Bel-7402 Cells and Its Resistant Variants by Activation of NOX4 and SIRT3.
BioMed research international
Li Y,Wang W,Xu X,Sun S,Xu X,Qu XJ
Thu Feb 04 00:00:00 EST 2016
{2-[1-(3-Methoxycarbonylmethyl-1H-indol-2-yl)-1-methyl-ethyl]-1H-indol-3-yl}-acetic Acid Methyl Ester Inhibited Hepatocellular Carcinoma Growth in Bel-7402 Cells and Its Resistant Variants by Activation of NOX4 and SIRT3.
BioMed research international
Li Y,Wang W,Xu X,Sun S,Xu X,Qu XJ
Thu Feb 04 00:00:00 EST 2016
Hydrogen sulfide improves the endothelial dysfunction in renovascular hypertensive rats.
Physiological research
Xue H,Zhou S,Xiao L,Guo Q,Liu S,Wu Y
Wed Sep 28 00:00:00 EDT 2016
Hydrogen sulfide improves the endothelial dysfunction in renovascular hypertensive rats.
Physiological research
Xue H,Zhou S,Xiao L,Guo Q,Liu S,Wu Y
Wed Sep 28 00:00:00 EDT 2016
Ubiquitin C-terminal hydrolase-L1 increases cancer cell invasion by modulating hydrogen peroxide generated via NADPH oxidase 4.
Oncotarget
Kim HJ,Magesh V,Lee JJ,Kim S,Knaus UG,Lee KJ
Tue Jun 30 00:00:00 EDT 2015
Ubiquitin C-terminal hydrolase-L1 increases cancer cell invasion by modulating hydrogen peroxide generated via NADPH oxidase 4.
Oncotarget
Kim HJ,Magesh V,Lee JJ,Kim S,Knaus UG,Lee KJ
Tue Jun 30 00:00:00 EDT 2015
Luteolin inhibits smooth muscle cell migration and proliferation by attenuating the production of Nox4, p-Akt and VEGF in endothelial cells.
Current pharmaceutical biotechnology
Ding H,Li D,Zhang Y,Zhang T,Zhu H,Xu T,Luo Y,Wang C
Mon Jan 05 00:00:00 EST 2015
Naringin inhibits TNF-α induced oxidative stress and inflammatory response in HUVECs via Nox4/NF-κ B and PI3K/Akt pathways.
Current pharmaceutical biotechnology
Li W,Wang C,Peng J,Liang J,Jin Y,Liu Q,Meng Q,Liu K,Sun H
Mon Aug 31 00:00:00 EDT 2015
NADPH oxidase 4 is required for interleukin-1β-mediated activation of protein kinase Cδ and downstream activation of c-jun N-terminal kinase signaling in smooth muscle.
Free radical biology & medicine
Ginnan R,Jourd'heuil FL,Guikema B,Simons M,Singer HA,Jourd'heuil D
Tue Jan 01 00:00:00 EST 2013
NADPH oxidase 4 is required for interleukin-1β-mediated activation of protein kinase Cδ and downstream activation of c-jun N-terminal kinase signaling in smooth muscle.
Free radical biology & medicine
Ginnan R,Jourd'heuil FL,Guikema B,Simons M,Singer HA,Jourd'heuil D
Tue Jan 01 00:00:00 EST 2013
Salvianolic acid A inhibits angiotensin II-induced proliferation of human umbilical vein endothelial cells by attenuating the production of ROS.
Acta pharmacologica Sinica
Yang LL,Li DY,Zhang YB,Zhu MY,Chen D,Xu TD
Sun Jan 01 00:00:00 EST 2012
Salvianolic acid A inhibits angiotensin II-induced proliferation of human umbilical vein endothelial cells by attenuating the production of ROS.
Acta pharmacologica Sinica
Yang LL,Li DY,Zhang YB,Zhu MY,Chen D,Xu TD
Sun Jan 01 00:00:00 EST 2012