Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
RSRP1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arginine/serine-rich protein 1; DJ465N24.2.1; RP3-465N24.4; UPF0471 protein C1orf63; UPF0471 protein C1orf63 homolog
Gene Aliases: 2700043I21Rik; C1orf63; D4Ucla2; D4Wsu53e; HT033; MNCb-0169; NPD014; RGD1359529; RSRP1
UniProt ID: (Human) Q9BUV0, (Mouse) Q3UC65, (Rat) Q5U2S0
Entrez Gene ID: (Human) 57035, (Mouse) 27981, (Rat) 362626
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support