Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody can recognize isoform 1, isoform 2, and isoform 3 of NPFFR2.
Receptor for NPAF (A-18-F-amide) and NPFF (F-8-F-amide) neuropeptides, also known as morphine-modulating peptides. Can also be activated by a variety of naturally occurring or synthetic FMRF-amide like ligands. This receptor mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 74; G-protein coupled receptor 74; G-protein coupled receptor HLWAR77; neuropeptide FF 2; Neuropeptide FF receptor 2; Neuropeptide G-protein coupled receptor; NPFF-R2; NPFF2 receptor; NPGP receptor
Gene Aliases: GPR74; HLWAR77; NPFF2; NPFFR2; NPGPR
UniProt ID: (Human) Q9Y5X5
Entrez Gene ID: (Human) 10886
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support