Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalyzes the cleavage of N-acetylneuraminic acid (sialic acid) to form pyruvate and N-acetylmannosamine via a Schiff base intermediate. It prevents sialic acids from being recycled and returning to the cell surface. Involved in the N- glycolylneuraminic acid (Neu5Gc) degradation pathway. Although human is not able to catalyze formation of Neu5Gc due to the inactive CMAHP enzyme, Neu5Gc is present in food and must be degraded.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dihydrodipicolinate synthetase homolog 1; N-acetylneuraminate lyase; N-acetylneuraminate pyruvate lyase (dihydrodipicolinate synthase); N-acetylneuraminate pyruvate-lyase; N-acetylneuraminic acid aldolase; NALase; Sialate lyase; Sialate-pyruvate lyase; Sialic acid aldolase; Sialic acid lyase
Gene Aliases: 0610033B02Rik; C112; C1orf13; NAL; NPL; NPL1
UniProt ID: (Human) Q9BXD5, (Mouse) Q9DCJ9
Entrez Gene ID: (Human) 80896, (Mouse) 74091
Molecular Function:
lyase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support