Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuroplastin; SDR-1; stromal cell derived factor receptor 1; Stromal cell-derived receptor 1
Gene Aliases: AW554172; GP55; GP65; np55; np65; NPTN; SDFR1; SDR-1; SDR1
UniProt ID: (Human) Q9Y639, (Mouse) P97300
Entrez Gene ID: (Human) 27020, (Dog) 487637, (Mouse) 20320
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support