Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NPTX1 is a member of the neuronal pentraxin family. Human neuronal pentraxin 1 is similar to rat NP1 which is a binding protein for the snake venom toxin taipoxin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 47 kDa taipoxin-binding protein; neuronal pentraxin 1; Neuronal pentraxin I; Neuronal pentraxin-1; NP-I; NP1
Gene Aliases: NP1; NPTX1
UniProt ID: (Human) Q15818, (Rat) P47971
Entrez Gene ID: (Human) 4884, (Rat) 266777
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support