Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Neuropeptide Y receptors are a class of G-protein coupled receptors which are activated by the closely related peptide horomones neuropeptide Y, peptide YY and pancreatic polypeptide. These receptors are involved in control of a diverse set of behavioral processes including appetite, circadian rhythme, and anxiety.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FC5; Neuropepetide Y1 Receptor; neuropeptide Y receptor subtype; Neuropeptide Y receptor type 1; neuropeptide Y1 receptor; NPY-1 receptor; NPY-Y1 receptor; NPY1-R; PY receptor Y1
Gene Aliases: NPY-1; Npy1r; Npyr; Y1-R
UniProt ID: (Mouse) Q04573
Entrez Gene ID: (Mouse) 18166, (Rat) 29358
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support