Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MEVNAGGVIA YISSSSSASS HASCHSEGSE NSFQSSSSSV PSSPNSSNSD TNGNPKNGDL ANIEGILKND RIDCSMKTSK SSAPGMTKSH SGVTKFSGMV LLCKVCGDVA SGFHYGVHAC EGCKGFFRRS IQQNIQYKKC LKNENCSIMR MNRNRCQQCR FKKCLSVGMS RDAVRFGRIP KREKQRMLIE MQSAMKTMMN SQFSGHLQND TLVEHHEQTA LPAQEQLRPK PQLEQENIKS SSPPSSDFAK EEVIGMVTRA HKDTFMYNQE QQENSAESMQ PKRGERIRKN MEQYNLNHDH CGNGLSSHFP CSESQQHLNG QFKGRNIMHY PNGHAICIAN GHCMNFSNAY TQRVCDRVPI DGFSQNENKN SYLCNTGGRM HLVCPMSKSP YVDPHKSGHE IWEEFSMSFT PAVKEVVEFA KRIPGFRDLS QHDQVNLLKA GTFEVLMVRF ASLFDAKERT VTFLSGKKYS VDDLHSMGAG DLLNSMFEFS EKLNALQLSD EEMSLFTAVV LVSADRSGIE NVNSVEALQE TLIRALRTLI MKNHPNEASI FTKLLLKLPD LRSLNNMHSE ELLAFKVHP
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EAR-1R; HZF2; nuclear receptor Rev-ErbA beta variant 1; nuclear receptor Rev-ErbA beta variant 2; Nuclear receptor subfamily 1 group D member 2; nuclear receptor subfamily 1, group D, member 2; Orphan nuclear hormone receptor BD73; Rev-erb alpha-related receptor; Rev-erb-beta; Rev-erba beta; rev-erba-alpha-related receptor; Rev-erbabeta; RVR; V-erbA-related protein 1-related
Gene Aliases: BD73; EAR-1R; NR1D2; RVR
UniProt ID: (Human) Q14995
Entrez Gene ID: (Human) 9975
Molecular Function:
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support