Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPHSAGGTAG VGLEAAEPTA LLTRAEPPSE PTEIRPQKRK KGPAPKMLGN ELCSVCGDKA SGFHYNVLSC EGCKGFFRRS VIKGAHYICH SGGHCPMDTY MRRKCQECRL RKCRQAGMRE ECVLSEEQIR LKKLKRQEEE QAHATSLPPR ASSPPQILPQ LSPEQLGMIE KLVAAQQQCN RRSFSDRLRV TPWPMAPDPH SREARQQRFA HFTELAIVSV QEIVDFAKQL PGFLQLSRED QIALLKTSAI EVMLLETSRR YNPGSESITF LKDFSYNRED FAKAGLQVEF INPIFEFSRA MNELQLNDAE FALLIAISIF SADRPNVQDQ LQVERLQHTY VEALHAYVSI HHPHDRLMFP RMLMKLVSLR TLSSVHSEQV FALRLQDKKL PPLLSEIWDV HE
NR1H3 (oxysterols receptor LXR-alpha) is a nuclear receptor that exhibits a ligand-dependent transcriptional activation activity. Its interaction with retinoic acid receptor (RXR) shifts RXR from its role as a silent DNA-binding partner to an active ligand-binding subunit in mediating retinoid responses through target genes defined by LXRES. LXRES are DR4-type response elements characterized by direct repeats of two similar hexanucleotide half-sites spaced by four nucleotides. NR1H3 plays an important role in the regulation of cholesterol homeostasis, regulating cholesterol uptake through MYLIP-dependent ubiquitination of LDLR, VLDLR, and LRP8. It also interplays functionally with RORA for the regulation of genes involves in liver metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: liver X nuclear receptor alpha variant 1; Liver X receptor alpha; LXRb; NER; NERI; NR1H2; Nuclear receptor subfamily 1 group H member 3; nuclear receptor subfamily 1, group H, member 3; Oxysterols receptor LXR-alpha
Gene Aliases: LXR-a; LXRA; NR1H3; RLD-1
UniProt ID: (Human) Q13133
Entrez Gene ID: (Human) 10062
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support