Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Immunogen sequence: MAMVVSTWRD PQDEVPGSQG SQASQAPPVP GPPPGAPHTP QTPGQGGPAS TPAQTAAGGQ GGPGGPGSDK QQQQQHIECV VCGDKSSGKH YGQFTCEGCK; Positive Samples: MCF-7, 293T; Cellular Location: Nucleus
This gene encodes a member of the steroid thyroid hormone superfamily of nuclear receptors. The encoded protein is a ligand inducible transcription factor that is involved in the regulation of many different genes. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor related protein 1; Apolipoprotein A-I regulatory protein 1; Apolipoprotein AI regulatory protein 1; apolipoprotein regulatory protein 1; ARP-1; chicken ovalbumin; chicken ovalbumin upstream promoter transcription factor 2; chicken ovalbumin upstream promoter-transcription factor I; COUP transcription factor 2; COUP transcription factor II; COUP-TF II; COUP-TF2; COUPb; MGC117452; Nuclear receptor subfamily 2 group F member 2; nuclear receptor subfamily 2, group F, member 2; Ovalbumin upstream promoter beta nuclear receptor
Gene Aliases: 2700033K02Rik; 9430015G03Rik; Aporp1; ARP-1; ARP1; CHTD4; COUP-TF2; COUP-TFII; COUPTFB; COUPTFII; EAR3; NF-E3; NR2F1; NR2F2; SVP40; Tcfcoup2; TFCOUP2
UniProt ID: (Human) P24468, (Rat) O09018, (Mouse) P43135
Entrez Gene ID: (Human) 7026, (Rat) 113984, (Mouse) 11819
Molecular Function:
C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support