Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control:HeLa cells.
Cellular Location: Nucleus.
Steroid receptors are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Mineralocorticoids are a family of steroids, secreted by the adrenal cortex, necessary for the regulation of a number of metabolic processes including electrolyte regulation. These compounds exert their effect through their interaction with the mineralocorticoid receptor (MR) and that complex's subsequent association with DNA. Given the function of mineralocorticoids, it is not surprising to find that the kidney is a primary target organ for mineralocorticoids and that this organ has been shown to contain MR. The corresponding gene for the mineralocorticoid receptor is NR3C2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aldosterone receptor; Mineralocorticoid receptor; mineralocorticoid receptor 1; mineralocorticoid receptor 2; mineralocorticoid receptor delta; MR; Nuclear receptor subfamily 3 group C member 2; nuclear receptor subfamily 3, group C, member 2 variant 3
Gene Aliases: MCR; MLR; MR; NR3C2; NR3C2VIT
UniProt ID: (Human) P08235
Entrez Gene ID: (Human) 4306
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support