Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: GFQVQHSPMW DDPGSLHNFH QNYVATTHMI EQRKTPVSRL SLFSFKQSPP GTPVSSCQMR FDGPLHVPMN PEPAGSHHVV DGQTFAVPNP IRKPASMGFP
The Nuclear receptor related 1 protein (Nurr1) also known as NR4A2 (nuclear receptor subfamily 4, group A, member 2) is a protein that in humans is encoded by the NR4A2 gene. NURR1 is a member of the nuclear receptor family of intracellular transcription factors.Nurr1 plays a key role in the maintenance of the dopaminergic system of the brain. Mutations in this gene have been associated with disorders related to dopaminergic dysfunction, including Parkinson's disease, schizophrenia, and manic depression. Misregulation of this gene may be associated with rheumatoid arthritis. Four transcript variants encoding four distinct isoforms have been identified for this gene. Additional alternate splice variants may exist, but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HZF3; Immediate-early response protein NOT; intermediate-early receptor protein; NGFI-B beta; NGFI-B/nur77 beta-type transcription factor homolog; nuclear receptor related 1; Nuclear receptor subfamily 4 group A member 2; nuclear receptor subfamily 4, group A, member 2; nur related protein-1, human homolog of; orphan nuclear receptor NR4A2; Orphan nuclear receptor NURR1; T-cell nuclear receptor NOT; transcriptionally inducible nuclear receptor related 1; Transcriptionally-inducible nuclear receptor
Gene Aliases: HZF-3; NOT; NR4A2; NURR1; RNR1; TINUR
UniProt ID: (Human) P43354
Entrez Gene ID: (Human) 4929
Molecular Function: C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support