Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDYSYDEDL DELCPVCGDK VSGYHYGLLT CESCKGFFKR TVQNNKHYTC TESQSCKIDK TQRKRCPFCR FQKCLTVGMR LEAVRADRMR GGRNKFGPMY KRDRALKQQK KAQIRANGFK LETGPPMGVP PPPPPAPDYV LPPSLHGPEP KGLAAGPPAG PLGDFGAPAL PMAVPGAHGP LAGYLYPAFP GRAIKSEYPE PYASPPQPGL PYGYPEPFSG GPNVPELILQ LLQLEPDEDQ VRARILGCLQ EPTKSRPDQP AAFGLLCRMA DQTFISIVDW ARRCMVFKEL EVADQMTLLQ NCWSELLVFD HIYRQVQHGK EGSILLVTGQ EVELTTVATQ AGSLLHSLVL (1-349 aa encoded by BC032501)
Steroidogenic factor-1 (SF-1) is an orphan nuclear receptor involved in organ development and differentiation of the endocrine system. SF-1 has been shown to be essential for fetal development of the hypothalamus, pituitary gland, gonads, and the adrenal gland. SF-1 has been shown to promote transcriptional activity of many steroidogenic enzymes from the cytochrome P450 family. SF-1 contains two activation function domains, designated AF1 and AF2, common to many nuclear steroid receptors which have been shown to be necessary for interaction with other nuclear receptors as well as coactivators and corepressors including steroid receptor coactivator-1 (SRC-1), glucocorticoid receptor interacting protein 1 (GRIP1), cAMP response element binding protein (CREB) binding protein (CBP) and silencing mediator of retinoid acid and thyroid hormone receptor (SMRT).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adrenal 4 binding protein; Adrenal 4 binding protein (AD4BP); Adrenal 4-binding protein; ELP; Embryonal long terminal repeat-binding protein; Embryonal LTR-binding protein; fushi tarazu 1 factor homolog; Fushi tarazu factor homolog 1; Fushi tarazu factor homolog 1 (FTZ1); nuclear receptor AdBP4; Nuclear receptor subfamily 5 group A member 1; Nuclear receptor subfamily 5 group A member 1 (NR5A1); nuclear receptor subfamily 5, group A, member 1; SF-1; Steroid hormone receptor Ad4BP; Steroid hydroxylase positive regulator; Steroidogenic factor 1; Steroidogenic factor 1 (SF1); steroidogenic factor 1 nuclear receptor; steroidogenic factor-1; STF-1; STF1
Gene Aliases: AD4BP; Ad4BP/SF-1; ELP; ELP-3; Ftz-F1; FTZ1; FTZF1; hSF-1; NR5A1; POF7; SF-1; SF1; SPGF8; SRXY3
UniProt ID: (Human) Q13285, (Mouse) P33242, (Rat) P50569
Entrez Gene ID: (Human) 2516, (Mouse) 26423, (Rat) 83826
Molecular Function:
C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support