Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LAHQQSRRAD RLLAAGKYEE AISCHKKAAA YLSEAMKLTQ SEQAHLSLEL QRDSHMKQLL LIQERWKRAQ REERLKAQQN TDKDAAAHLQ TSHKPSAEDA EGQSPLSQKY SPSTEKCLPE IQGIFDRDPD TLLYLLQQKS EPAEPCIGSK APKDDKTIIE EQATKIADLK RHVEFLVAEN ERLRKENKQL KAEKARLLKG PIEKELDVDA D; Positive Samples: HepG2, 293T, HeLa, Jurkat; Cellular Location: Cytoplasm, Cytoplasmic vesicle, Nucleus, autophagosome
NRBF2 (nuclear receptor-binding factor 2) modulates transcriptional activation by target nuclear receptors. It is involved in starvation-induced autophagy probably by its association with PI3K complex I (PI3KC3-C1). NRBF2 stabilizes the PI3KC3-C1 assembly and enhances ATG14-linked lipid kinase activity of PIK3C3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Comodulator of PPAR and RXR; comodulator of PPAR and RXR 1; comodulator of PPAR and RXR 2; NRBF-2; nuclear receptor binding factor-2; Nuclear receptor-binding factor 2
Gene Aliases: COPR; COPR1; COPR2; NRBF-2; NRBF2
UniProt ID: (Human) Q96F24, (Rat) Q9QYK3, (Mouse) Q8VCQ3
Entrez Gene ID: (Human) 29982, (Rat) 58839, (Mouse) 641340
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support