Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PLMNFAATR PLGLPRVLAP PPEEVQKAKT PTPEPFDSET RKVIQMQCNL ERSEDKARWH LTLLLVLEDR LHRQLTYDLL PTDSAQDLAS ELVHYGFLHE DDRMKLAAFL ESTFLKYRGT QA (244-501 aa encoded by BC113873)
Nrbp2 may regulate apoptosis of neural progenitor cells during their differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NRBP2; Nuclear receptor-binding protein 2; Transformation-related gene 16 protein; transformation-related protein 16; TRG-16
Gene Aliases: BC011468; NRBP2; PP9320; TRG16
UniProt ID: (Human) Q9NSY0
Entrez Gene ID: (Human) 340371, (Pig) 110260412, (Mouse) 223649, (Rat) 680451
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support