Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: LVPSQTVVQT FSNPDGTVSL IQVGTGATVA TLADASELPT TVTVAQVNYS AVADGEVEQN WATLQGGEMT IQTTQASEAT QAVASLAEAA VAASQEMQQG A
This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for "nuclear factor (erythroid-derived 2)-like 1" which has an official symbol of NFE2L1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha palindromic-binding protein; Alpha-pal; EWG; NRF-1; Nuclear respiratory factor 1; nuclear respiratory factor-1
Gene Aliases: ALPHA-PAL; C87038; D6Ertd415e; Nfe2l1_retired; NRF1
UniProt ID: (Human) Q16656, (Mouse) Q9WU00
Entrez Gene ID: (Human) 4899, (Rat) 312195, (Mouse) 18181
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support