Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
FIGURE: 1 / 1
Immunogen sequence: SILSTEDPNQ LTVNSLNSDA TVNTDFGDEF YSAFIAEPSI SNSMPSPATL SHSLSELLNG PIDVSDLSLC KAFNQNHPES TAEFNDSDSG ISLNTSPSVA SPEHSVESSS YGDTLLGLSD SEVEELDSAP GSVKQNGPKT PVHSSGDMVQ PLSPSQGQST HVHDAQCENT PEKELPVSPG HRKTPFTKDK HSSRLEAHLT RDELRAKALH IPFPVEKIIN LPVVDFNEMM SKEQFNEAQL ALIRDIRRRG KNKVAAQNCR KRKLENIVEL EQDLDHLKDE KEKLLKEKGE NDKSLHLLKK QLSTLYLEVF SMLRDEDGKP YSPSEYSLQQ TRDGNVFLVP KSKKPDVKKN
Nfr1 (also designated TCF11), Nrf2 and Nfr3 are bZIP transcription factors that heterodimer with Maf proteins to bind MARE (Maf recognition elements) sequences. Nrf2 is widely expressed and is thought to translocate to the nucleus after treatment with xenobiotics and antioxidants, which stimulate its release from a repressor protein Keap1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E4TF1-60; E4TF1A; HEBP1; NF-E2-related factor 2; NFE2-related factor 2; NFT2; NRF2A; Nuclear factor erythroid 2-related factor 2; nuclear factor erythroid-derived 2-like 2; nuclear factor, erythroid 2-like 2; Nuclear factor, erythroid derived 2, like 2
Gene Aliases: AI194320; HEBP1; NFE2L2; NRF2
UniProt ID: (Human) Q16236, (Mouse) Q3UQK0
Entrez Gene ID: (Human) 4780, (Mouse) 18024
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support