Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: SLKSVQDQAY KAPVVVEGKV QGLVPAGGSS SNSTREPPAS GRVALVKVLD KWPLRSGGLQ REQVISVGSC VPLERNQRYI FFLEPTEQPL VFKTAFAPLD
Neuregulin 2 is a novel member of the neuregulin family of growth and differentiation factors. Through interaction with the Erbb family of receptors, NRG2 induces the growth and differentiation of epithelial, neuronal, glial, and other types of cells. The gene consists of 12 exons and the genomic structure is similar to that of neuregulin 1, another member of the neuregulin family of ligands. NRG1 and NRG2 mediate distinct biological processes by acting at different sites in tissues and eliciting different biological responses in cells. The gene is located close to the region for demyelinating Charcot-Marie-Tooth disease locus, but is not responsible for this disease. Alternative transcripts encoding distinct isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: divergent of neuregulin-1; neural- and thymus-derived activator for ErbB kinases; Pro-neuregulin-2, membrane-bound isoform; Pro-NRG2
Gene Aliases: DON1; HRG2; NRG2; NTAK
UniProt ID: (Human) O14511
Entrez Gene ID: (Human) 9542
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support