Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: 293T, HepG2, HeLa, MCF7, Mouse liver, Mouse brain, Mouse spleen, Mouse kidney, Mouse lung
Immunogen sequence: EEALSQAKKS VASKTEVKKT RRPRSVLNTQ PEQTNAGEVA SSLSSTEIRR HSQRRHTSAE EEEPPPVKIA WKTAAARKSL PASITMHKGS LDLQKCNMSP VVKIEQVFAL QNATGDGKFI DQFVYSTKGI GNKTEISVRG QDRLIISTPN QRNEKPTQSV SSPEATSGST GSVEKKQQRR SIRTRSESEK STEVVPKKKI K
This gene is related to the Wolf-Hirschhorn syndrome candidate-1 gene and encodes a protein with PWWP (proline-tryptophan-tryptophan-proline) domains. The function of the protein has not been determined. Two alternatively spliced variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp667H044; FLJ20353; Histone-lysine N-methyltransferase NSD3; MGC126766; MGC142029; nuclear receptor binding SET domain protein 3; Nuclear SET domain-containing protein 3; Protein whistle; WHSC1-like 1 isoform 9 with methyltransferase activity to lysine; WHSC1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like protein 1 homolog
Gene Aliases: 6720429E03; A530023P05; AI528490; DC28; NSD3; pp14328; WHISTLE; WHSC1L1
UniProt ID: (Human) Q9BZ95, (Mouse) Q6P2L6
Entrez Gene ID: (Human) 54904, (Rat) 290831, (Mouse) 234135
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support