Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5' UTR have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bare patches; NAD(P)-dependent steroid dehydrogenase-like protein; Protein H105e3; short chain dehydrogenase/reductase family 31E, member 1; Sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating; striated
Gene Aliases: AI747449; Bpa; H105E3; NSDHL; SDR31E1; Str; XAP104
UniProt ID: (Human) Q15738, (Mouse) Q9R1J0, (Rat) Q5PPL3
Entrez Gene ID: (Human) 50814, (Mouse) 18194, (Rat) 309262
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support