Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kinetochore-associated protein NSL1 homolog; NSL1, MIND kinetochore complex component, homolog
Gene Aliases: C1orf48; DC31; DC8; MIS14; NSL1
UniProt ID: (Human) Q96IY1
Entrez Gene ID: (Human) 25936
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support