Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RNEDLLEVG SRPGPASQLP RFVRVNTLKT CSDDVVDYFK RQGFSYQGRA SSLDDLRALK GKHFLLDPLM PELLVFPAQT DLHEHPLYRA GHLILQDRAS CLPAMLLDPP PGSHVIDACA APGNKTSHLA ALLKNQGKIF AFDLDAKRLA SMATLLARAG VSCCELAEED FLAVSPSDPR YHEVHYILLD PSCSGSGMPS RQLEEPGAGT PSPVRLHALA GFQQRALCHA LTFPSLQRLV YSTCSLCQEE NEDVVRDALQ QNPGAFRLAP ALPAWPHRGL STFPGAEHCL RASPETTLSS GFFVAVIERV EVPSSASQAK ASAPERTPSP APKRKKRQQR AAAGACTPPC T (117-466 aa encoded by BC008084)
NSUN5 is a member of the evolutionarily conserved NOL1/NOP2/Sun domain family. The encoded protein may function as a DNA methyltransferase in the nucleus. The gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S rRNA (cytosine-C(5))-methyltransferase; FLJ10267; MGC986; NOL1-related protein; NOL1/NOP2/Sun domain family member 5; NOL1/NOP2/Sun domain family, member 5; NOL1R; NOP2/Sun domain family, member 5; NOP2/Sun domain family, member 5A; probable 28S rRNA (cytosine-C(5))-methyltransferase; putative methyltransferase NSUN5; Williams Beuren syndrome chromosome region 20A; Williams Beuren syndrome chromosome region 20A homolog; Williams-Beuren syndrome chromosomal region 20A protein; Williams-Beuren syndrome chromosomal region 20A protein homolog; Williams-Beuren syndrome critical region protein 20; Williams-Beuren syndrome critical region protein 20 copy A
Gene Aliases: 9830109N13Rik; AI326939; NOL1; NOL1R; NSUN5; NSUN5A; p120; p120(NOL1); RGD1309268; WBSCR20; WBSCR20A
UniProt ID: (Human) Q96P11, (Mouse) Q8K4F6
Entrez Gene ID: (Human) 55695, (Mouse) 100609, (Rat) 288595
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support