Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSTSWSDRLQ NAADMPANMD KHALKKYRRE AYHRVFVNRS LAMEKIKCFG FDMDYTLAVY KSPEYESLGF ELTVERLVSI GYPQELLSFA YDSTFPTRGL VFDTLYGNLL KVDAYGNLLV CAHGFNFIRG PETREQYPNK FIQRDDTERF YILNTLFNLP ETYLLACLVD FFTNCPRYTS CETGFKDGDL FMSYRSMFQD VRDAVDWVHY KGSLKEKTVE NLEKYVVKDG KLPLLLSRMK EVGKVFLATN SDYKYTDKIM TYLFDFPHGP KPGSSHRPWQ SYFDLILVDA RKPLFFGEGT VLRQVDTKTG KLKIGTYTGP LQHGIVYSGG SSDTICDLLG AKGKDILYIG DHIFGDILKS KKRQGWRTFL VIPELAQELH VWTDKSSLFE ELQSLDIFLA ELYKHLDSSS NERPDISSIQ RRIKKVTHDM DMCYGMMGSL FRSGSRQTLF ASQVMRYADL YAASFINLLY YPFSYLFRAA HVLMPHESTV EHTHVDINEM ESPLATRNRT SVDFKDTDYK RHQLTRSISE IKPPNLFPLA PQEITHCHDE DDDEEEEEEE E
Purine 5-prime-nucleotidase (EC 3.1.3.5) preferentially hydrolyzes inosine 5-prime-monophosphate (IMP) and other purine nucleotides, and is allosterically activated by various compounds, including ATP. The enzyme is exclusively located in the cytoplasmic matrix of cells and may have a critical role in the maintenance of a constant composition of intracellular purine/pyrimidine nucleotides in cooperation with other nucleotidases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-nucleotidase (purine), cytosolic type B; cN-II; Cytosolic 5'-nucleotidase II; Cytosolic IMP/GMP-specific 5'-nucleotidase; Cytosolic nucleoside phosphotransferase 5'N; Cytosolic purine 5'-nucleotidase; High Km 5'-nucleotidase; IMP-specific 5'-NT; RP11-30H12.3
Gene Aliases: cN-II; GMP; NT5B; NT5C2; NT5CP; PNT5; SPG45; SPG65
UniProt ID: (Human) P49902
Entrez Gene ID: (Human) 22978
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support