Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTNQESAVH VKMMPEFQKS SVRIKNPTRV EEIICGLIKG GAAKLQIITD FDMTLSRFSY KGKRCPTCHN IIDNCKLVTD ECRKKLLQLK EKYYAIEVDP VLTVEEKYPY MVEWYTKSHG LLVQQALPKA KLKEIVAESD VMLKEGYENF FDKLQQHSIP VFIFSAGIGD VLEEVIRQAG VYHPNVKVVS NFMDFDETGV LKGFKGELIH VFNKHDGALR NTEYFNQLKD NSNIILLGDS QGDLRMADGV ANVEHILKIG YLNDRVDELL EKYMDSYDIV LVQDESLEVA NSILQKIL (1-297 aa encoded by BC015856)
Pyrimidine 5-prime-nucleotidase (P5N), also called uridine 5-prime monophosphate hydrolase (UMPH), catalyzes the dephosphorylation of the pyrimidine 5-prime monophosphates UMP and CMP to the corresponding nucleosides. There are 2 isozymes of pyrimidine 5-prime nucleotidase in red blood cells, referred to as type I (UMPH1) and type II (UMPH2). The 2 enzymes are not separable by electrophoresis in humans but have distinct kinetic properties, and the proteins show no homology.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-nucleotidase, cytosolic III; 5'-nucleotidase, pyrimidine; 7-methylguanosine nucleotidase; 7-methylguanosine phosphate-specific 5'-nucleotidase; cN-III; Cytosolic 5'-nucleotidase 3; Cytosolic 5'-nucleotidase 3A; Cytosolic 5'-nucleotidase III; Lupin; MGC27337; MGC87109; MGC87828; p36; P5'N-1; P5N-1; Pyrimidine 5'-nucleotidase 1; Uridine 5'-monophosphate hydrolase 1; uridine monophosphatase 1
Gene Aliases: 1600024P05Rik; 2610206B05Rik; 3110004A18Rik; AI842776; cN-III; HSPC233; hUMP1; lupin; NT5C3; NT5C3A; Nt5y; p36; P5'N-1; P5N-1; P5N1; PN-1; PN-I; POMP; PSN1; UMPH; Umph-1; UMPH1
UniProt ID: (Human) Q9H0P0, (Mouse) Q9D020
Entrez Gene ID: (Human) 51251, (Mouse) 107569, (Rat) 312373
Molecular Function:
esterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support