Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The MW of this protein is 14 kDa, and this antibody specifically recognizes the 14 kDa protein.
Immunogen sequence: IGSSFIQHY YQLFDNDRTQ LGAIYIDASC LTWEGQQFQG KAAIVEKLSS LPFQKIQHSI TAQDHQPTPD SCIISMVVGQ LKADEDPIMG FHQMFLLKNI NDAWVCTNDM FRLALHNFG (10-127 aa encoded by BC002348)
The protein encoded by this gene is a cytosolic factor that facilitates protein transport into the nucleus. It interacts with the nuclear pore complex glycoprotein p62. This encoded protein acts at a relative late stage of nuclear protein import, subsequent to the initial docking of nuclear import ligand at the nuclear envelope. It is thought to be part of a multicomponent system of cytosolic factors that assemble at the pore complex during nuclear import.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NTF-2; Nuclear transport factor 2; Placental protein 15; PP15
Gene Aliases: NTF-2; NTF2; NUTF2; PP15
UniProt ID: (Human) P61970
Entrez Gene ID: (Human) 10204
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support