Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NYREN18 is a specific down-regulator of the NEDD8 conjugation system. It recruits NEDD8 and its conjugates to the proteasome for degradation. It also interacts with PSMD4/S5a, a member of the regulatory subunit of the 26S proteasome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NEDD8 ultimate buster 1; NEDD8 ultimate buster-1; Negative regulator of ubiquitin-like proteins 1; Renal carcinoma antigen NY-REN-18
Gene Aliases: BS4; NUB1; NUB1L; NYREN18
UniProt ID: (Human) Q9Y5A7
Entrez Gene ID: (Human) 51667
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support