Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEAAAEPGN LAGVRHIILV LSGKGGVGKS TISTELALAL RHAGKKVGIL DVDLCGPSIP RMLGAQGRAV HQCDRGWAPV FLDREQSISL MSVGFLLEKP DEAVVWRGPK KNALIKQFVS DVAWGELDYL VVDTPPGTSD EHMATIEALR PYQPLGALVV TTPQAVSVGD VRRELTFCRK TGLRVMGIVE NMSGFTCPHC TECTSVFSRG GGEELAQLAG VPFLGSVPLD PALMRTLEEG HDFIQEFPGS PAFAALTSIA QKILDATPAC LP (1-271 aa encoded by BC002768)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytosolic Fe-S cluster assembly factor NUBP2; cytosolic Fe-S cluster assembly factor NUBP2 {ECO:0000255|HAMAP-Rule:MF_03039}; homolog of yeast cytosolic Fe-S cluster deficient 1; NBP 2; NBP 2 {ECO:0000255|HAMAP-Rule:MF_03039}; nucleotide binding protein 2 (MinD homolog, E. coli); Nucleotide-binding protein 2; nucleotide-binding protein 2 {ECO:0000255|HAMAP-Rule:MF_03039}
Gene Aliases: CFD1; D17Wsu11e; NBP 2; NUBP1; NUBP2
UniProt ID: (Human) Q9Y5Y2, (Mouse) Q9R061, (Rat) Q68FS1
Entrez Gene ID: (Human) 10101, (Mouse) 26426, (Rat) 287125
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support