Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSVKRSLK QEIVTQFHCS AAEGDIAKLT GILSHSPSLL NETSENGWTA LMYAARNGHP EIVQFLLEKG CDRSIVNKSR QTALDIAVFW GYKHIANLLA TAKGGKKPWF LTNEVEECEN YFSKTLLDRK SEKRNNSDWL LAKESHPATV FILFSDLNPL VTLGGNKESF QQPEVRLCQL NYTDIKDYLA QPEKITLIFL GVELEIKDKL LNYAGEVPRE EEDGLVAWFA LGIDPIAAEE FKQRHENCYF LHPPMPALLQ LKEKEAGVVA QARSVLAWHS RYKFCPTCGN ATKIEEGGYK RLCLKEDCPS LNGVHNTSYP RVDPVVIMQV IHPDGTKCLL GRQKRFPPGM FTCLAGFIEP GETIEDAVRR EVEEESGV (1-377 aa encoded by BC041099)
Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DeNADding enzyme NUDT12; DKFZP761I172; NAD-capped RNA hydrolase NUDT12; NADH pyrophosphatase NUDT12; nucleoside diphosphate linked moiety X-type motif 12; Nucleoside diphosphate-linked moiety X motif 12; nudix (nucleoside diphosphate linked moiety X)-type motif 12; nudix (nucleoside diphosphnudix (nucleoside diphosphate linked moiety X)-type motif 12ate linked moiety X)-type motif 12; Nudix motif 12; nudix-type motif 12; peroxisomal NADH pyrophosphatase NUDT12
Gene Aliases: 0610016O18Rik; NUDT12
UniProt ID: (Human) Q9BQG2, (Mouse) Q9DCN1
Entrez Gene ID: (Human) 83594, (Rat) 367323, (Mouse) 67993
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support